Skip to Content
Merck
All Photos(1)

Key Documents

N-046

Supelco

(+)-Norpseudoephedrine hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Cathine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14ClNO
CAS Number:
Molecular Weight:
187.67
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.C[C@H](N)[C@@H](O)c1ccccc1

InChI

1S/C9H13NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7,9,11H,10H2,1H3;1H/t7-,9+;/m0./s1

InChI key

DYWNLSQWJMTVGJ-DKXTVVGFSA-N

General description

A certified reference solution suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications, from clinical toxicology and forensic analysis to urine drug testing and sports testing. (+)-Norpseudoephedrine, also known as cathine, is a psychoactive stimulant of the phenethylamine and amphetamine classes.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuei Hui Chan et al.
Journal of analytical toxicology, 33(3), 162-166 (2009-04-18)
Sho-seiryu-to is one of the most common Traditional Chinese Medicine preparations for the attenuation of colds. Ephedrae Herba is one of the prescriptions of Sho-seiryu-to. The major ingredients of Ephedrae Herba, ephedrines, are banned substances on the World Anti-Doping Agency
Raz Krizevski et al.
Journal of ethnopharmacology, 114(3), 432-438 (2007-10-12)
Khat (Catha edulis Forsk., Celastraceae) is a perennial shrub that was introduced to Israel by Yemenite immigrants. Khat young leaves are chewed as a stimulant. The main stimulating active principles in this plant are the phenylpropylamino alkaloids (-)-cathinone [(S)-alpha-aminopropiophenone], (+)-cathine
Hesham Abdul Aziz et al.
The journal of sexual medicine, 6(3), 682-695 (2009-01-16)
Khat (Catha edulis) is an evergreen tree/shrub that is thought to affect sexual motivation or libido. Its positive effect on sexual desire is more frequently observed in females than in males and occurs when khat is chewed. Thus, khat's effects
Osquel Barroso et al.
Drug testing and analysis, 4(5), 320-329 (2011-05-07)
Due to its stimulatory effects on the central nervous system, and its structural similarity to banned stimulants such as ephedrine and methamphetamine, pseudoephedrine (PSE) at high doses is considered as an ergogenic aid for boosting athletic performance. However, the status
K Deventer et al.
Drug testing and analysis, 1(5), 209-213 (2010-04-01)
Until the end of 2003 a urinary concentration of pseudoephedrine exceeding 25 microg/mL was regarded as a doping violation by the World Anti-Doping Agency. Since its removal from the prohibited list in 2004 the number of urine samples in which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service