Skip to Content
Merck
All Photos(1)

Key Documents

M-167

Supelco

5-MeO-DiPT

1.0 mg/mL in methanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26N2O
CAS Number:
Molecular Weight:
274.40
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[nH]1c2c(c(c1)CCN(C(C)C)C(C)C)cc(cc2)OC

InChI

1S/C17H26N2O/c1-12(2)19(13(3)4)9-8-14-11-18-17-7-6-15(20-5)10-16(14)17/h6-7,10-13,18H,8-9H2,1-5H3

InChI key

DNBPMBJFRRVTSJ-UHFFFAOYSA-N

General description

5-MeO-DiPT (5-methoxy-diisopropyltryptamine), known by the street names Foxy or Foxy Methoxy, is a designer tryptamine that can cause auditory distortions and hallucinations. Reports suggest that 5-MeO-DiPT is seven times more potent than the hallucinogen DMT as a result of the methoxy substitution at position five on the tryptamine ring. This Certified Spiking Solution® is suitable for use in a variety of GC/MS or LC/MS testing applications in forensic toxicology, urine drug testing, or clinical chemistry.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service