Skip to Content
Merck
All Photos(1)

Key Documents

M-084

Supelco

Modafinil solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO2S
CAS Number:
Molecular Weight:
273.35
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

NC(=O)CS(=O)C(c1ccccc1)c2ccccc2

InChI

1S/C15H15NO2S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17)

InChI key

YFGHCGITMMYXAQ-UHFFFAOYSA-N

Gene Information

human ... SLC6A3(6531)

General description

An analeptic drug approved by the US FDA for the treatment of narcolepsy, shift work sleep disorder, and excessive daytime sleepiness. This well known nootropic drug, also known as Provigil® or Modavigil®, is often used as a performance-enhancing drug in academics and sports.

Application


  • Innovative Spectrofluorimetric Method for Modafinil Determination: An innovative computationally designed-spectrofluorimetric method has been developed for the determination of modafinil in tablets and human plasma, leveraging the high-purity modafinil solution. This approach enhances analytical accuracy and is crucial for pharmacokinetic studies and neuropharmacological research, offering significant benefits for biochemistry experiments focused on cognitive enhancement and neurotransmitter studies (Hosny et al., 2022).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Modavigil is a registered trademark of Cephalon, Inc.
Provigil is a registered trademark of Cephalon, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trends in on-label and off-label modafinil use in a nationally representative sample.
Renée A Peñaloza et al.
JAMA internal medicine, 173(8), 704-706 (2013-04-05)
Mariana Raineri et al.
PloS one, 7(10), e46599-e46599 (2012-10-12)
Methamphetamine is a drug of abuse that can cause neurotoxic damage in humans and animals. Modafinil, a wake-promoting compound approved for the treatment of sleeping disorders, is being prescribed off label for the treatment of methamphetamine dependence. The aim of
Jared Bruce et al.
Cognitive and behavioral neurology : official journal of the Society for Behavioral and Cognitive Neurology, 25(3), 107-114 (2012-09-11)
Examine the efficacy of armodafinil in improving cognition in patients with multiple sclerosis (MS). Many patients with MS experience cognitive difficulties. Armodafinil has shown promise as a cognitive enhancer in other patient populations. No studies have examined whether armodafinil improves
Mei-Hong Qiu et al.
PloS one, 7(9), e45471-e45471 (2012-10-03)
We have previously shown that modafinil promotes wakefulness via dopamine receptor D(1) and D(2) receptors; however, the locus where dopamine acts has not been identified. We proposed that the nucleus accumbens (NAc) that receives the ventral tegmental area dopamine inputs
Cognitive enhancers are set to change workplaces.
Jacqui Wise
BMJ (Clinical research ed.), 345, e7538-e7538 (2012-11-09)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service