Skip to Content
Merck
All Photos(1)

Key Documents

H-026

Supelco

(±)-11-Hydroxy-Δ9-THC solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC(CCCCC)=CC2=C1C3C=C(CO)CCC3C(O2)(C)C

InChI

1S/C21H30O3/c1-4-5-6-7-14-11-18(23)20-16-10-15(13-22)8-9-17(16)21(2,3)24-19(20)12-14/h10-12,16-17,22-23H,4-9,13H2,1-3H3

InChI key

YCBKSSAWEUDACY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® applicable in LC/MS or GC/MS applications for cannabinoid testing, urine drug testing, clinical toxicology, or forensic analysis. 11-Hydroxy-Δ9-THC is an active metabolite of THC and metabolic precursor to 11-nor-9-carboxy-Δ9-THC. This cannabinoid is found in biological matrices including urine, blood, and plasma.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Del Mar Ramirez Fernandez et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 875(2), 465-470 (2008-10-17)
Cannabis is considered to be the most widely abused illicit drug in Europe. Consequently, sensitive and specific analytical methods are needed for forensic purposes and for cannabinoid pharmacokinetic and pharmacodynamic studies. A simple, rapid and highly sensitive and specific method
Cynthia Coulter et al.
Journal of analytical toxicology, 32(8), 653-658 (2008-11-15)
An analytical procedure for the determination of Delta9-tetrahydrocannabinol (THC), 11-nor-9-carboxy-Delta9-tetrahydrocannabinol (THCA), and 11-hydroxy-Delta9-tetrahydrocannabinol (11-OH-THC) in whole blood has been developed and validated using liquid chromatography with tandem mass spectral detection (MS). Cannabinoids present in the blood samples were quantified using
Erin L Karschner et al.
Clinical chemistry, 57(1), 66-75 (2010-11-17)
Sativex(®), a cannabis extract oromucosal spray containing Δ(9)-tetrahydrocannabinol (THC) and cannabidiol (CBD), is currently in phase III trials as an adjunct to opioids for cancer pain treatment, and recently received United Kingdom approval for treatment of spasticity. There are indications
Erin L Karschner et al.
Analytical and bioanalytical chemistry, 397(2), 603-611 (2010-03-23)
A sensitive analytical method for simultaneous quantification of sub-nanogram concentrations of cannabidiol (CBD), Delta(9)-tetrahydrocannabinol (THC), 11-hydroxy-THC (11-OH-THC), and 11-nor-9-carboxy-THC (THCCOOH) in plasma is presented for monitoring cannabinoid pharmacotherapy and illicit cannabis use. Analytes were extracted from 1 mL plasma by
J Röhrich et al.
Journal of analytical toxicology, 34(4), 196-203 (2010-05-15)
Cannabinoid concentrations in blood and urine after passive exposure to cannabis smoke under real-life conditions were investigated in this study. Eight healthy volunteers were exposed to cannabis smoke for 3 h in a well-attended coffee shop in Maastricht, Netherlands. An

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service