Skip to Content
Merck
All Photos(1)

Key Documents

F-002

Supelco

Fentanyl solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N2O
CAS Number:
Molecular Weight:
336.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CCC(=O)N(C1CCN(CC1)CCc2ccccc2)c3ccccc3

InChI

1S/C22H28N2O/c1-2-22(25)24(20-11-7-4-8-12-20)21-14-17-23(18-15-21)16-13-19-9-5-3-6-10-19/h3-12,21H,2,13-18H2,1H3

InChI key

PJMPHNIQZUBGLI-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

A potent synthetic narcotic analgesic and local anesthetic with a rapid onset and short duration of action. Sold under various trade names including Sublimaze, Duragesic®, Actiq®, and Fentora®, fentanyl is more potent than morphine in analgesic activity. This certified solution standard is suitable for a variety of LC/MS or GC/MS testing applications from forensic analysis, clinical toxicology, and pain prescription monitoring to urine drug testing and pharmaceutical research.

Application


  • Analgesic potency assay: Fentanyl is widely used in analgesic potency assays due to its strong opioid receptor agonist properties, providing a benchmark for evaluating the efficacy of new analgesics in preclinical and clinical settings (Chakravartula et al., 2024).

  • In vitro pharmacology studies: As a high-purity fentanyl analytical standard, it is crucial for in vitro pharmacology studies to understand the drug′s interaction with opioid receptors and its effects on neural pathways (Kanashiro et al., 2024).

  • Pharmacological research chemical supply: Fentanyl citrate solution serves as a valuable resource in pharmacological research, particularly in studies focusing on pain management and opioid addiction therapies (Lee et al., 2024).

  • Synthetic opioid solution: This synthetic opioid is utilized in scientific studies to explore safer and more effective pain relief methods, such as patient-controlled analgesia systems (Huang et al., 2024).

  • Opioid receptor agonist research: Fentanyl is employed to study opioid receptor dynamics, aiding in the development of targeted therapies for pain and opioid use disorders (Song et al., 2024).

Legal Information

Actiq is a registered trademark of Anesta Corp.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Duragesic is a registered trademark of Johnson & Johnson
Fentora is a registered trademark of Cima Labs, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nele Plock et al.
International journal of clinical pharmacology and therapeutics, 51(6), 495-508 (2013-04-25)
Intranasal Fentanyl Spray (INFS) was developed for the treatment of breakthrough pain (BTP) in cancer patients using a new route of administration. Dose strengths of 50, 100, and 200 μg INFS (Instanyl®) are currently on the market, however, some adult
An EMS guide to chronic pain.
Kevin T Collopy et al.
EMS world, 42(7), 59-63 (2013-08-10)
[Pain therapy. Treating pain patients with opioid preparations].
MMW Fortschritte der Medizin, 155(6), 70-71 (2013-05-25)
Esther Encinas et al.
Paediatric drugs, 15(3), 247-257 (2013-05-10)
Fentanyl is a synthetic opioid commonly used as an anesthetic and also increasingly popular as a sedative agent in neonates. Initial dosage regimens in this population are often empirically derived from adults on a body weight basis. However, ontogenic maturation
Hong-Xia Zhu et al.
Zhongguo zhen jiu = Chinese acupuncture & moxibustion, 33(4), 342-346 (2013-07-04)
To observe the clinical effect of acupoint injection with small dose of fentanyl-droperido mixed liquor at different time stages on labor analgesia, and explore its function mechanism. One hundred and fifty cases of full-term primiparas who were intended to take

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service