Skip to Content
Merck

Skip To

E-021

EDDP-D3 perchlorate solution

100 μg/mL in methanol (as pyrrolinium), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

EDDP-D3 perchlorate solution, Methadone metabolite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20D3H21N · ClO4
CAS Number:
Molecular Weight:
380.88
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as pyrrolinium)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=Cl(=O)([O-])=O.C[N+]1=C(CC([2H])([2H])[2H])C(C2=CC=CC=C2)(C3=CC=CC=C3)CC1C

InChI

1S/C20H24N.ClHO4/c1-4-19-20(15-16(2)21(19)3,17-11-7-5-8-12-17)18-13-9-6-10-14-18;2-1(3,4)5/h5-14,16H,4,15H2,1-3H3;(H,2,3,4,5)/q+1;/p-1/i1D3;

InChI key

FICCBMGBBWWZRF-NIIDSAIPSA-M

General description

EDDP is the major urinary metabolite of methadone, a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. This stable-labeled internal standard is suitable for EDDP and methadone testing methods by GC/MS or LC/MS for forensic analysis, urine drug testing or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E-062E-022E-006
description

100 μg/mL in methanol (as pyrrolinium), ampule of 1 mL, certified reference material, Cerilliant®

description

1.0 mg/mL in methanol (as pyrrolinium), ampule of 1 mL, certified reference material, Cerilliant®

description

1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®

description

ampule of 10 mg, certified reference material, Cerilliant®

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

application(s)

forensics and toxicology

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

solid

format

single component solution

format

single component solution

format

single component solution

format

neat

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material


Still not finding the right product?

Explore all of our products under EDDP-D3 perchlorate solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters