Skip to Content
Merck

D-164

N-Desmethylmirtazapine-D4 solution

100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H13D4N3
CAS Number:
Molecular Weight:
255.35
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

concentration

100 μg/mL in acetonitrile

format

single component solution

storage temp.

−20°C

General description

A stable-labled internal standard suitable for quantitation of N-Desmethylmirtazapine levels in urine, serum, or plasma by LC/MS or GC/MS for forensic analysis, clinical toxicology, or urine drug testing. N-Desmethylmirtazapine is the major urinary metabolite of the tetracyclic antidepressant mirtazapine, marketed under the trade names Remeron®, Avanza, or Zispin for treatment of depression. The metabolite, also known as normirtazapine, is pharmacologically active.

Legal Information

Remeron is a registered trademark of MSD
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library