Skip to Content
Merck
All Photos(1)

Documents

D-045

Supelco

Doxylamine succinate solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28N2O5
CAS Number:
Molecular Weight:
388.46
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

OC(=O)CCC(O)=O.CN(C)CCOC(C)(c1ccccc1)c2ccccn2

InChI

1S/C17H22N2O.C4H6O4/c1-17(20-14-13-19(2)3,15-9-5-4-6-10-15)16-11-7-8-12-18-16;5-3(6)1-2-4(7)8/h4-12H,13-14H2,1-3H3;1-2H2,(H,5,6)(H,7,8)

InChI key

KBAUFVUYFNWQFM-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

Doxylamine is a member of the ethanolamine class of antihistamines and is the most effective over-the-counter (OTC) sedative in the US. This Snap-N-Spike® Reference Solution is applicable to use in LC/MS or GC/MS applications for clinical toxicology or forensic analysis.

Application


  • Quality assessment of doxylamine succinate: A chemometric approach was applied for quality assessment of doxylamine succinate with its degradation product, using predictive models based on UV-spectrophotometric data, highlighting its utility in ensuring the integrity of pharmaceutical formulations (Abd El-Hadi et al., 2022).

  • Analysis in over-the-counter medications: Doxylamine succinate was analyzed as part of a quaternary mixture in cold medications, utilizing a sequential spectrophotometric resolution approach, indicating its role in multi-component pharmaceutical analysis (Kelani et al., 2023).

  • LC-MS/MS method for plasma levels: A sensitive LC-MS/MS method was developed for determining doxylamine levels in human plasma, demonstrating its application in pharmacokinetic studies and bioequivalence assessments (Fu et al., 2022).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Eccles et al.
The Journal of pharmacy and pharmacology, 47(12A), 990-993 (1995-12-01)
Antihistamines are widely used in common cold medications, although the role of histamine in the development of common cold symptoms is unclear and the use of antihistamines for the treatment of common cold is controversial. It is clear that antihistamines
OTC product: Zicam Cold and Flu Medicated Spoons.
Julie L Olenak
Journal of the American Pharmacists Association : JAPhA, 46(6), e52-e53 (2007-05-23)
B Leybishkis et al.
The American journal of the medical sciences, 322(1), 48-49 (2001-07-24)
Doxylamine succinate, an over-the-counter antihistamine, is commonly used as a nighttime sleep aid in the short-term management of insomnia. It is also used in combination with antitussive and decongestant agents for the temporary relief of common cold symptoms. Doxylamine is
C L Holder et al.
Journal of analytical toxicology, 14(4), 247-251 (1990-07-11)
Elimination and metabolic profiles of the glucuronide products of doxylamine and its N-demethylated metabolites were determined after the oral administration of (14C)-doxylamine succinate (13.3 and 133 mg/kg doses) to male and female Fischer 344 rats. The cumulative urinary and fecal
L G Roberts et al.
Toxicology and applied pharmacology, 97(1), 134-140 (1989-01-01)
The intracellular pH of the early postimplantation rodent embryo (pHi) is alkaline with respect to the corresponding plasma of the pregnant dam. This transplacental pH gradient is of considerable importance in the accumulation of teratogenic weak acids by the embryo.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service