Skip to Content
Merck
All Photos(1)

Key Documents

C-019

Supelco

S(-)-Cathinone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12ClNO
CAS Number:
Molecular Weight:
185.65
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.C[C@H](N)C(=O)c1ccccc1

InChI

1S/C9H11NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7H,10H2,1H3;1H/t7-;/m0./s1

InChI key

HPZCAKYHISJOIK-FJXQXJEOSA-N

General description

This Certified Spiking Solution® is suitable for GC/MS or LC/MS testing applications involving cathinone in clinical toxicology, forensic analysis or urine drug testing. Cathinone, or benzoylethanamine, is a major active constituent of the recreational drug “Khat”. This stimulant has also been found in illicit bath salts.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laureen J Marinetti et al.
Journal of analytical toxicology, 37(3), 135-146 (2013-01-31)
To date, the Toxicology Section of the Montgomery County Coroner's Office/Miami Valley Regional Crime Laboratory has identified six synthetic cathinones, commonly found in bath salt products, in 43 cases. Thirty-two cases will be reviewed here, including all of the postmortem
Michael H Baumann et al.
European journal of pharmacology, 698(1-3), 1-5 (2012-11-28)
Recently there has been a dramatic rise in the abuse of so-called "bath salts" products that are purchased as legal alternatives to illicit drugs like cocaine and 3,4-methylenedioxymethamphetamine (MDMA). Baths salts contain one or more synthetic derivatives of the naturally-occurring
Psychoactive 'bath salts': compounds, mechanisms, and toxicities.
Kurt R Lehner et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 38(1), 243-244 (2012-11-14)
Przemysław Biliński et al.
Annals of agricultural and environmental medicine : AAEM, 19(4), 857-870 (2013-01-15)
Faced with the rapidly growing increase of designer drug abuse, particularly amongst the younger generation, various legislative strategies are currently employed world-wide for tackling this problem - however with mixed results. The key issue is that the producers of DDs
S E Shortall et al.
British journal of pharmacology, 168(4), 966-977 (2012-10-10)
Recreational users report that mephedrone has similar psychoactive effects to 3,4-methylenedioxymethamphetamine (MDMA). MDMA induces well-characterized changes in body temperature due to complex monoaminergic effects on central thermoregulation, peripheral blood flow and thermogenesis, but there are little preclinical data on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service