Skip to Content
Merck
All Photos(1)

Documents

B-908

Supelco

Buprenorphine-D4 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37NO4D4
CAS Number:
Molecular Weight:
471.66
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1([2H])CC1C([2H])([2H])N2CC[C@]34[C@H]5Oc6c(O)ccc(C[C@@H]2[C@]37CC[C@@]5(OC)[C@H](C7)[C@](C)(O)C(C)(C)C)c46

InChI

1S/C29H41NO4/c1-25(2,3)26(4,32)20-15-27-10-11-29(20,33-5)24-28(27)12-13-30(16-17-6-7-17)21(27)14-18-8-9-19(31)23(34-24)22(18)28/h8-9,17,20-21,24,31-32H,6-7,10-16H2,1-5H3/t20-,21-,24-,26+,27-,28+,29-/m1/s1/i6D2,16D2/t17?,20-,21-,24-,26+,27-,28+,29-

InChI key

RMRJXGBAOAMLHD-UHGBJPCQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An internal standard suitable for buprenorphine testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, pain prescription monitoring, urine drug testing, or forensic analysis. Buprenorphine is a semi-synthetic opiate used in the treatment of moderate acute pain and opioid addiction. The drug is sold under numerous trade names such as Suboxone®, Subutex®, and Temgesic.

Application


  • HPLC-MS for sildenafil citrate analysis: Buprenorphine-d4 is used as the internal standard (IS) in the liquid-liquid extraction procedure for the determination of sildenafil in biological fluids by HPLC-MS. Tracqui and Ludes, 2003.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Suboxone is a registered trademark of RB Pharmaceuticals Limited
Subutex is a registered trademark of RB Pharmaceuticals Limited

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service