Skip to Content

B-030

Butalbital-D5 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Butalbital-D5 solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11D5H11N2O3
CAS Number:
Molecular Weight:
229.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

N1C(=O)NC(=O)C(C1=O)(CCC(C)C)C(C(=C([2H])[2H])[2H])([2H])[2H]

InChI

1S/C12H18N2O3/c1-4-6-12(7-5-8(2)3)9(15)13-11(17)14-10(12)16/h4,8H,1,5-7H2,2-3H3,(H2,13,14,15,16,17)/i1D2,4D,6D2

InChI key

OFGPEYHYGSLQGU-LDEUCTAFSA-N

General description

A certified analytical reference solution and internal standard applicable for use in butalbital testing methods by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing or forensic analysis. Butalbital is a barbiturate often combined with other medications such as acetaminophen or aspirin for the treatment of pain or codeine for headaches. Trade names of several butalbital combinations include Axocet®, Bucet, and Bupap® with acetaminophen and Axotal with aspirin.

Legal Information

Axocet is a registered trademark of Fougera Pharmaceuticals Inc.
Bupap is a registered trademark of ECR Pharmaceuticals Co., Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Butalbital-D5 solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library