Skip to Content
Merck
All Photos(2)

Key Documents

900407P

Avanti

16:0-pacFA PC

Avanti Research - A Croda Brand 900407P, powder

Synonym(s):

1-palmitoyl-2-(9-(3-pent-4-ynyl-3-H-diazirin-3-yl)-nonanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72N3O8P
CAS Number:
Molecular Weight:
741.98
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1x10 mg (900407P-10MG)
pkg of 1 × 1 mg (900407P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900407P

lipid type

click reagents
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C#CCCCC1(N=N1)CCCCCCCCC(O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C39H72N3O8P/c1-6-8-10-11-12-13-14-15-16-17-18-21-24-28-37(43)47-34-36(35-49-51(45,46)48-33-32-42(3,4)5)50-38(44)29-25-22-19-20-23-27-31-39(40-41-39)30-26-9-7-2/h2,36H,6,8-35H2,1,3-5H3/t36-/m1/s1

InChI key

XBLDRLHMTZEHJM-PSXMRANNSA-N

General description

The pacFA lipid attached to 1-palmitoyl-sn-glycero-3-phosphocholine, contains a photoactivable diazirine ring with a clickable alkyne group.

Application

1-palmitoyl-2-(9-(3-pent-4-ynyl-3-H-diazirin-3-yl)-nonanoyl)-sn-glycero-3-phosphocholine (16:0-pacFA PC) might be used to label hydrogenated zwitterionic bicelles (HZB) and hydrogenated anionic bicelles (HAB).

Biochem/physiol Actions

1-palmitoyl-2-(9-(3-pent-4-ynyl-3-H-diazirin-3-yl)-nonanoyl)-sn-glycero-3-phosphocholine (16:0-pacFA PC) may be used for a global analysis of protein–lipid interactions in living cells. pacFA acts as a precursor for the biosynthesis of bifunctional lipids. Proteins in contact with the bifunctional lipids are then cross-linked by ultraviolet (UV) irradiation of the diazirine ring. Finally, click chemistry is used to label the alkyne with a reporter molecule.

Packaging

5 mL Amber Glass Screw Cap Vial (900407P-10MG)
5 mL Amber Glass Screw Cap Vial (900407P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael Dos Santos Morais et al.
Biophysical journal, 115(7), 1231-1239 (2018-09-11)
Scaffolding proteins play important roles in supporting the plasma membrane (sarcolemma) of muscle cells. Among them, dystrophin strengthens the sarcolemma through protein-lipid interactions, and its absence due to gene mutations leads to the severe Duchenne muscular dystrophy. Most of the
Dominique Mias-Lucquin et al.
Journal of structural biology, 209(1), 107411-107411 (2019-11-07)
Dystrophin is a large intracellular protein that prevents sarcolemmal ruptures by providing a mechanical link between the intracellular actin cytoskeleton and the transmembrane dystroglycan complex. Dystrophin deficiency leads to the severe muscle wasting disease Duchenne Muscular Dystrophy and the milder
Per Haberkant et al.
Biochimica et biophysica acta, 1841(8), 1022-1030 (2014-01-21)
Understanding biological processes at the mechanistic level requires a systematic charting of the physical and functional links between all cellular components. While protein-protein and protein-nucleic acid networks have been subject to many global surveys, other critical cellular components such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service