890813P
Avanti
VSA 2
Synonym(s):
Semisynthetic Saponin Adjuvant-VSA 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C94H147NO42
CAS Number:
Molecular Weight:
1963.15
MDL number:
UNSPSC Code:
12352211
Recommended Products
Assay
≥99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (890813P-1MG)
pkg of 1 × 5 mg (890813P-5MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
application(s)
vaccine development
shipped in
dry ice
storage temp.
−20°C
General description
VSA-2 is a semisynthetic saponin adjuvant derived from the naturally available Momordica saponin, designed to stimulate antigen-specific humoral and cellular immune responses. It is created by adding a terminal-functionalized side chain to the branched trisaccharide domain at the C3 position of Momordica saponin II (MS II), which is extracted from the seeds of Momordica cochinchinensis Spreng. When compared directly to the clinically validated saponin adjuvant QS-21, VSA-2 demonstrates comparable effectiveness in enhancing both humoral and cellular immune responses.
Application
Vaccine development
Packaging
5 mL Amber Glass Screw Cap Vial (890813P-1MG); 5 mL Amber Glass Screw Cap Vial (890813P-5MG)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service