Skip to Content
Merck
All Photos(1)

Key Documents

878114P

Avanti

C18-02:0 PC

Avanti Research - A Croda Brand 878114P, powder

Synonym(s):

1-O-octadecyl-2-acetyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H58NO7P
CAS Number:
Molecular Weight:
551.74
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (878114P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878114P

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C)=O)COCCCCCCCCCCCCCCCCCC)=O

InChI

1S/C28H58NO7P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-23-33-25-28(36-27(2)30)26-35-37(31,32)34-24-22-29(3,4)5/h28H,6-26H2,1-5H3/t28-/m1/s1

InChI key

ZXCIEWBDUAPBJF-MUUNZHRXSA-N

General description

C18-02:0 PC, also referred as 1-O-octadecyl-2-acetyl-sn-glycero-3-phosphocholine, is a natural platelet-activating factor. It is found in variety of cells including, basophil leucocytes, macrophages, polymorphonuclear neutrophils and monocytes and platelets.

Application

C18-02:0 PC or 1-O-octadecyl-2-acetyl-sn-glycero-3-phosphocholine may be used as a substrate in enzymatic lipoprotein-associated phospholipase A2 (Lp-PLA2) activity assay.

Biochem/physiol Actions

C18-02:0 PC acts as a platelet-activating factor and facilitates platelet aggregation and secretion. It also plays a key role in inflammation and anaphylaxis.

Packaging

5 mL Clear Glass Sealed Ampule (878114P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott D Ryan et al.
Journal of lipid research, 49(10), 2250-2258 (2008-06-14)
The platelet-activating factor (PAF) family of glycerophospholipids accumulates in damaged brain tissue following injury. Little is known about the role of individual isoforms in regulating neuronal survival. Here, we compared the neurotoxic and neuroprotective activities of 1-O-hexadecyl-2-acetyl-sn-glycero-3-phosphocholine (C16-PAF) and 1-O-octadecyl-2-acetyl-sn-glycero-3-phosphocholine
Saki Yamaura et al.
Clinica chimica acta; international journal of clinical chemistry, 481, 184-188 (2018-03-20)
Measurement of lipoprotein-associated phospholipase A2 (Lp-PLA2) can be used as an adjunct to traditional cardiovascular risk factors for identifying individuals at higher risk of cardiovascular events. This can be performed by quantification of the protein concentration using an ELISA platform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service