Skip to Content
Merck
All Photos(1)

Key Documents

860811P

Avanti

12:0(2R-OH) Ceramide

Avanti Research - A Croda Brand 860811P, powder

Synonym(s):

N-(2′-(R)-hydroxylauroyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H59NO4
CAS Number:
Molecular Weight:
497.79
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 5 mg (860811P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860811P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@H](O)CCCCCCCCCC)=O)CO

InChI

1S/C30H59NO4/c1-3-5-7-9-11-13-14-15-16-17-19-20-22-24-28(33)27(26-32)31-30(35)29(34)25-23-21-18-12-10-8-6-4-2/h22,24,27-29,32-34H,3-21,23,25-26H2,1-2H3,(H,31,35)/b24-22+/t27-,28+,29+/m0/s1

InChI key

CPDJNFOAARXMMT-XOVVVQEXSA-N

General description

Ceramides containing 2-hydroxy fatty acid (hFA) are found primarily in the epidermis.[1] 12:0(2R-OH) Ceramide is a 2R hydroxylated lauric acid-containing ceramide.[2]

Biochem/physiol Actions

Hydroxy fatty acid (hFA)-sphingolipids play a specific role in cell signaling. Synthesis of hFA-ceramides requires fatty acid 2-hydroxylase (FA2H). Mutations of this key enzyme are associated with the development of nervous system disorders such as leukodystrophy and spastic paraparesis in humans. hFA-ceramides are essential for the epidermal permeability barrier function.[3] The mechanism of action for an antitumor drug, PM02734 involves hFA-ceramides in the cell membrane.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (860811P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levels of SCS7/FA2H-mediated fatty acid 2-hydroxylation determine the sensitivity of cells to antitumor PM02734
Herrero AB, et al.
Cancer research, 68(23), 9779-9787 (2008)
Normal fur development and sebum production depends on fatty acid 2-hydroxylase expression in sebaceous glands
Maier H, et al.
The Journal of Biological Chemistry, 286(29), 25922-25934 (2011)
Helena Maier et al.
The Journal of biological chemistry, 286(29), 25922-25934 (2011-06-02)
2-Hydroxylated fatty acid (HFA)-containing sphingolipids are abundant in mammalian skin and are believed to play a role in the formation of the epidermal barrier. Fatty acid 2-hydroxylase (FA2H), required for the synthesis of 2-hydroxylated sphingolipids in various organs, is highly
Fatty acid 2-hydroxylase regulates cAMP-induced cell cycle exit in D6P2T schwannoma cells
Alderson NL, et al.
Journal of Lipid Research, 50(6), 1203-1208 (2009)
Fatty acid 2-Hydroxylation in mammalian sphingolipid biology
Hama H, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1801(4), 405-414 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service