Skip to Content
Merck
All Photos(1)

Key Documents

860658P

Avanti

sphinganine-d7

Avanti Research - A Croda Brand 860658P, powder

Synonym(s):

D-erythro-sphinganine-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32D7NO2
CAS Number:
Molecular Weight:
308.55
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860658P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860658P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@@](O)([H])CCCCCCCCCCCCC(C(C([2H])([2H])[2H])([2H])[2H])([2H])[2H]

General description

Sphinganine-d7 is a deuterated derivative of sphinganine. Sphinganine, also called as dihydrosphingosine is a derivative of sphingosine. Sphinganines are produced in endoplasmic reticulum, by decarboxylating condensation of palmitoyl-CoA and serine.

Application

Sphinganine-d7 has been used as an internal standard for quantification of sphingolipid concentrations in plasma and tissue samples by high-performance liquid chromatography tandem mass spectrometry.

Biochem/physiol Actions

Sphinganine (SPA) acts as a selective inhibitor of protein kinase C in intact human neutrophils. It may function a potential biomarker for the diagnosis of sphingolipidoses. SPA accumulation enables fumonisin B1 to stimulate apoptosis. It also has an ability to block cholesterol transport in Niemann-Pick Type C (NPC) disease.

Packaging

5 mL Amber Glass Screw Cap Vial (860658P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gennaro Bruno et al.
Cellular signalling, 45, 110-121 (2018-02-07)
Skeletal muscle tissue retains a remarkable regenerative capacity due to the activation of resident stem cells that in pathological conditions or after tissue damage proliferate and commit themselves into myoblasts. These immature myogenic cells undergo differentiation to generate new myofibers
Lipids
Basic neurochemistry: principles of molecular, cellular, and medical neurobiology (2011)
D Pittet et al.
The Journal of biological chemistry, 262(21), 10072-10076 (1987-07-25)
Sphinganine has been proposed to be a specific inhibitor of protein kinase C. In the present study we have evaluated whether sphinganine is a convenient tool to probe for the role of protein kinase C in neutrophil function. Human neutrophils
Federica Storti et al.
eLife, 8 (2019-03-14)
Age-related macular degeneration (AMD) is a progressive disease of the retinal pigment epithelium (RPE) and the retina leading to loss of central vision. Polymorphisms in genes involved in lipid metabolism, including the ATP-binding cassette transporter A1 (ABCA1), have been associated
Graham Brogden et al.
Biomolecules, 9(9) (2019-09-11)
Sphingolipids are an important group of lipids that play crucial roles in living cells, facilitating cell recognition, signal transduction and endocytosis. The concentration of sphingosine and some of its derivatives like sphinganine may serve as a biomarker for the diagnosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service