Skip to Content
Merck
All Photos(1)

Key Documents

860496O

Avanti

Dimethyl Sphingosine (d18:1)

N,N-dimethyl-D-erythro-sphingosine, neat oil

Synonym(s):

(2R,3S,E)-2-(dimethylamino)octadec-4-ene-1,3-diol, N,N-DMS, D-erythro-N,N-dimethylsphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H41NO2
CAS Number:
Molecular Weight:
327.55
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 5 mg (860496O-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860496O

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N(C)C)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C20H41NO2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)19(18-22)21(2)3/h16-17,19-20,22-23H,4-15,18H2,1-3H3/b17-16+/t19-,20+/m0/s1

InChI key

YRXOQXUDKDCXME-YIVRLKKSSA-N

General description

Dimethyl Sphingosine (d18:1), also called as N,N-Dimethyl-d-erythro-sphingosine (DMS), is a natural sphingolipid metabolite. It is found in various tissues.

Application

Dimethyl Sphingosine (d18:1) may be used:
  • to investigate D-erythro-N,N-dimethylsphingosine (DMS)-induced cell death in U937 cells to understand its molecular mechanism
  • to study its effects on intracellular Ca2+ concentration, pH and glutamate uptake in human 1321N1 astrocytes
  • in substrate specificity studies of sphingosine methyltransferase activity in rat kidney homogenate

Dimethyl sphingosine (d18:1) has been used as an inhibitor of sphingosine kinase.

Biochem/physiol Actions

D-erythro-N,N-dimethylsphingosine (DMS) prevents the activity of sphingosine kinase and protein kinase C. It modulates cellular activities. DMS exhibits anti-cancer and apoptotic properties. It is involved in the augmentation of epidermal growth factor receptor (EGFR) and inhibition of src kinase. DMS functions as a potential therapeutic for Chagas disease cardiomyopathy through its anti-parasitic and immunomodulatory properties.
D-erythro-N,N-dimethylsphingosine (DMS) has an ability to inhibit the activity of sphingosine kinase. It also helps to study the role of sphingosine 1-phosphate (SPP) as an intracellular second messenger. DMS exhibits anti-parasitic and immunomodulatory activities. It acts as a potential therapeutic for chronic Chagas disease cardiomyopathy.

Packaging

5 mL Clear Glass Sealed Ampule (860496O-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-throughput screening assay for sphingosine kinase inhibitors in whole blood using RapidFire mass spectrometry
Highkin MK, et al.
Journal of Biomolecular Screening, 16(2), 272-277 (2011)
Therapeutic effects of sphingosine kinase inhibitor N, N-dimethylsphingosine (DMS) in experimental chronic Chagas disease cardiomyopathy
Vasconcelos JF, et al.
Scientific reports, 7(1), 6171-6171 (2017)
N-(4-Hydroxyphenyl) retinamide increases dihydroceramide and synergizes with dimethylsphingosine to enhance cancer cell killing
Wang H, et al.
Molecular Cancer Therapeutics, 7(9), 2967-2976 (2008)
Hongtao Wang et al.
Molecular cancer therapeutics, 7(9), 2967-2976 (2008-09-16)
Fenretinide [N-(4-hydroxyphenyl)retinamide (4-HPR)] is cytotoxic in many cancer cell types. Studies have shown that elevation of ceramide species plays a role in 4-HPR cytotoxicity. To determine 4-HPR activity in a multidrug-resistant cancer cell line as well as to study ceramide
Hyo-Lim Kim et al.
Prostaglandins & other lipid mediators, 86(1-4), 18-25 (2008-03-05)
In the present study, we studied N,N-dimethyl-D-erythro-sphingosine (DMS)-induced cell death and its signaling mechanism in U937 human monocytes. We found that DMS induced cell death in a concentration-dependent manner, while sphingosine 1-phosphate did not. DMS also induced DNA fragmentation, nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service