Skip to Content
Merck
All Photos(2)

Key Documents

850360C

Avanti

17:0 PC

1,2-diheptadecanoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

PC(17:0/17:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H84NO8P
CAS Number:
Molecular Weight:
762.09
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850360C-25mg)
pkg of 2 × 20 mL (850360C-1g)

manufacturer/tradename

Avanti Research - A Croda Brand 850360C

concentration

10 mg/mL (850360C-25mg)
25 mg/mL (850360C-1g)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OCC[N+](C)(C)C)OCC(OC(=O)CCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCC

InChI

1S/C42H84NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-41(44)48-38-40(39-50-52(46,47)49-37-36-43(3,4)5)51-42(45)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h40H,6-39H2,1-5H3

InChI key

RTWAYAIMWLNAJW-UHFFFAOYSA-N

General description

17:0 PC is a saturated phospholipid.[1] Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes.[2] It is also an important component of the mucosal layer of the colon.[3]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

17:0 PC is suitable for use as an internal standard in the quantitation of lipid species in biological samples by liquid chromatography-mass spectrometry[4][5] and fatty acid analyses in gas chromatography.[6]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[3] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[7]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850360C-1g)
5 mL Clear Glass Sealed Ampule (850360C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stearoyl-CoA desaturase 1, elongase 6 and their fatty acid products and precursors are altered in ovariectomized rats with 17beta-estradiol and progesterone treatment
Marks KA, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 89(2-3), 89-96 (2013)
Phase transition of a single lipid bilayer measured by sum-frequency vibrational spectroscopy
Liu J and Conboy JC
Journal of the American Chemical Society, 126(29), 8894-8895 (2004)
The Membranes of Cells (2016)
Integrative Medicine - E-Book (2017)
Lipidomic analysis of endocannabinoid metabolism in biological samples
Astarita G and Piomelli D
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 877(26), 2755-2767 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service