Skip to Content
Merck
All Photos(3)

Key Documents

840054C

Avanti

Soy PC

Avanti Research - A Croda Brand 840054C

Synonym(s):

L-α-phosphatidylcholine (Soy)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840054C-25mg)
pkg of 2 × 4 mL (840054C-200mg)
pkg of 5 × 4 mL (840054C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840054C

concentration

10 mg/mL (840054C-25mg)
25 mg/mL (840054C-200mg)
25 mg/mL (840054C-500mg)

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

General description

L-α-phosphatidylcholine (Soy) is a naturally occurring phospholipid and the most abundant phospholipid in eukaryotic cellular membrane with high amount of PUFAs.

Application

Soy PC may be used for the synthesis of proteoliposomes, lipoprotein particle (LLP) mimics and flexible liposome (FLUV).

Biochem/physiol Actions

L-α-phosphatidylcholine plays an important role in the formation of liposomes. It is also used in small unilamellar vesicles (SUVs) synthesis.

Packaging

5 mL Clear Glass Sealed Ampule (840054C-200mg)
5 mL Clear Glass Sealed Ampule (840054C-25mg)
5 mL Clear Glass Sealed Ampule (840054C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piezo1 Channels Are Inherently Mechanosensitive
Syeda R, et al.
Testing, 17(7), 1739-1746 (2016)
Membrane curvature recognition by C reactive protein using lipoprotein mimics
Wang MS, et al.
Soft Matter, 8(30), 7909-7918 (2012)
Superresolution and fluorescence dynamics evidence reveal that intact liposomes do not cross the human skin barrier.
Dreier J, et al.
PLoS ONE, 11(1), e0146514-e0146514 (2016)
Louiza Bohn Thomsen et al.
Materials (Basel, Switzerland), 12(21) (2019-11-07)
Magnetic nanoparticles have great prospects for drug delivery purposes, as they can be designed with various surface coatings and conjugated with drugs and targeting moieties. They also have a unique potential for precise delivery when guided by magnetic force. The
María Cecilia Izquierdo et al.
Journal of cosmetic dermatology (2020-04-14)
Ultradeformable liposomes are promising carriers for cosmeceutical actives as they can be loaded with molecules of different polarities, and they present unique penetration properties. While those features have already been tested, we wanted to know whether their special penetration properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service