Skip to Content
Merck
All Photos(3)

Documents

840012P

Avanti

Heart CA

Avanti Research - A Croda Brand 840012P, powder

Synonym(s):

Cardiolipin (Heart, Bovine) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 mg (840012P-100mg)
pkg of 1 × 25 mg (840012P-25mg)
pkg of 1 × 50 mg (840012P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840012P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)OCC([H])(O)COP([O-])(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)=O.[Na+].[Na+]

InChI

1S/C81H142O17P2.2Na/c1-5-9-13-17-21-25-29-33-37-41-45-49-53-57-61-65-78(83)91-71-76(97-80(85)67-63-59-55-51-47-43-39-35-31-27-23-19-15-11-7-3)73-95-99(87,88)93-69-75(82)70-94-100(89,90)96-74-77(98-81(86)68-64-60-56-52-48-44-40-36-32-28-24-20-16-12-8-4)72-92-79(84)66-62-58-54-50-46-42-38-34-30-26-22-18-14-10-6-2;;/h21-28,33-40,75-77,82H,5-20,29-32,41-74H2,1-4H3,(H,87,88)(H,89,90);;/q;2*+1/p-2/b25-21-,26-22-,27-23-,28-24-,37-33-,38-34-,39-35-,40-36-;;/t76-,77-;;/m1../s1

InChI key

ZGSPNIOCEDOHGS-LBOJVHLDSA-L

General description

Cardiolipin is a unique phospholipid present in the inner mitochondrial membrane, which makes up to 20% of total lipids. It is a non-bilayer anionic phospholipid, which has four acyl chains and small headgroup.

Application

Heart CA has been used as a standard stock solution for its quantitative analysis using liquid chromatography−mass spectrometry (LC-MS)/MS. It has also been used for liposome preparation.

Biochem/physiol Actions

Cardiolipin is involved in the dynamic organization of membranes and in cell division. It is implicated in proton uptake pathway. Cardiolipin plays a vital role in mitochondrial biogenesis, fusion and fission, respiration and protein import. It regulates cell wall biogenesis, vacuole homeostasis, aging, cell cycle and apoptosis.

Packaging

5 mL Clear Glass Sealed Ampule (840012P-100mg)
5 mL Clear Glass Sealed Ampule (840012P-25mg)
5 mL Clear Glass Sealed Ampule (840012P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of cardiolipin in cardiovascular health
Shen Z, et al.
BioMed Research International, 2015(1) (2015)
Cryopreservation of lipid bilayers by LEA proteins from Artemia franciscana and trehalose
Moore DS and Hand SC
Cryobiology, 73(2), 240-247 (2016)
Role of cardiolipin in mitochondrial signaling pathways
Dudek J
Frontiers in Cell and Developmental Biology, 5(1), 90-90 (2017)
A rapid and sensitive LC-MS/MS method for quantitative analysis of cardiolipin (18: 2) 4 in human leukocytes and mouse skeletal muscles
Xu G, et al.
Journal of Pharmaceutical and Biomedical Analysis, 158(1), 386-394 (2018)
Hay-Yan J Wang et al.
Journal of the American Society for Mass Spectrometry, 18(3), 567-577 (2006-12-13)
Cardiolipins (CL) are mitochondria specific lipids. They play a critical role in ATP synthesis mediated by oxidative phosphorylation. Abnormal CL distribution is associated with several disease states. MALDI-MS and MALDI-MS/MS were used to demonstrate in situ analysis and characterization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service