Skip to Content
Merck
All Photos(1)

Key Documents

810209P

Avanti

C6-NBD Ceramide

Avanti Research - A Croda Brand

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H49N5O6
CAS Number:
Molecular Weight:
575.74
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810209P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C30H49N5O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-27(37)25(23-36)32-28(38)19-16-14-17-22-31-24-20-21-26(35(39)40)30-29(24)33-41-34-30/h15,18,20-21,25,27,31,36-37H,2-14,16-17,19,22-23H2,1H3,(H,32,38)/b18-15+/t25-,27+/m0/s1

InChI key

HZIRBXILQRLFIK-VPZZKNKNSA-N

General description

C6-NBD ceramide is a fluorescent labelled compound, known to be useful for the detection of enzymatic action. C6-NBD ceramide is preferred over radioactive compounds.[1]

Application

C6-NBD Ceramide is suitable as a substrate for sphingomyelin synthase activity assay.[2]

Biochem/physiol Actions

Ceramide is one of the products of sphingolipid metabolism. The members of ceramide synthases family are responsible for its synthesis. Each ceramide synthesized differs in its acyl chain length. Ceramides possess a wide range of functions, associated with apoptosis, cancer inhibition and chemotherapy. Its action is also related to neurodegenerative diseases.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810209P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Glycosyltransferases and Related Genes, 5-5 (2011)
A role for ceramides, but not sphingomyelins, as antagonists of insulin signaling and mitochondrial metabolism in C2C12 myotubes
Park M, et al.
The Journal of Biological Chemistry, 291(46), 23978-23988 (2016)
Mammalian ceramide synthases
Levy M and Futerman AH
IUBMB Life, 62(5), 347-356 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service