Skip to Content
Merck
All Photos(2)

Key Documents

810155C

Avanti

18:1-06:0 NBD PE

Avanti Research - A Croda Brand 810155C

Synonym(s):

1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H58N5O11P
CAS Number:
Molecular Weight:
755.84
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810155C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810155C

concentration

1 mg/mL (810155C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is a fluorescent probe where the fluorescent NBD label is attached to the headgroup of phosphatidylethanolamine. The NBD moiety is known to localize to the membrane interface. Phosphoethanolamine is the second most abundant andmetabolically related aminophospholipid. It is mostly enriched in brain.

Application

18:1-06:0 NBD PE has been used as a liposomal supplement for protein reconstitution into liposomes and scrambling experiments. It may also be used to analyse the temporal fluorescent pattern of Chol-E-NBD.

Biochem/physiol Actions

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) plays a vital role as a fluorescent probe in the studies of organization and dynamics of membranes. Phosphoethanolamine (PE) acts as a substrate for several phospholipids of the cell membrane.

Packaging

5 mL Amber Glass Screw Cap Vial (810155C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bieong-Kil Kim et al.
Bioorganic & medicinal chemistry letters, 25(18), 3893-3896 (2015-08-06)
Cholesterol-based fluorescent lipids with ether linker were synthesized using NBD (Chol-E-NBD) or Rhodamine B (Chol-E-Rh), and the usefulnesses as fluorescent probes for tracing cholesterol-based liposomes were validated. The fluorescent intensities of liposomes containing these modified lipids were measured and observed
Synthetic phosphoethanolamine induces cell cycle arrest and apoptosis in human breast cancer MCF-7 cells through the mitochondrial pathway
Ferreira AK, et al.
Biomedicine and Pharmacotherapy, 37-50 (2012)
Formation and function of phosphatidylserine and phosphatidylethanolamine in mammalian cells
Vance JE and Tasseva G
The FEBS journal, 37-50 (2012)
Fluorescent methods to study biological membranes, 37-50 (2012)
Organization and dynamics of N-(7-nitrobenz-2-oxa-1, 3-diazol-4-yl)-labeled lipids: a fluorescence approach
Mukherjee S, et al.
Chemistry and Physics of Lipids, 37-50 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service