Skip to Content
Merck
All Photos(1)

Key Documents

800811C

Avanti

18:1 DG

1-2-dioleoyl-sn-glycerol, chloroform

Synonym(s):

1-2-di-(9Z-octadecenoyl)-sn-glycerol; DG(18:1(9Z)/18:1(9Z)/0:0); DOG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O5
CAS Number:
Molecular Weight:
620.99
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (800811C-10mg)
pkg of 1 × 5 mL (800811C-25mg)
pkg of 1 × 8 mL (800811C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800811C

concentration

2 mg/mL (800811C-10mg)
25 mg/mL (800811C-200mg)
5 mg/mL (800811C-25mg)

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C39H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37,40H,3-16,21-36H2,1-2H3/b19-17+,20-18-/t37-/m0/s1

InChI key

AFSHUZFNMVJNKX-CSWDEDFYSA-N

General description

18:1 DG or 1,2-dioleoyl-sn-glycerol (DOG) comprises a small polar region and is synthesized via phosphatidic acid intermediate.
In biochemical signaling, diacylglycerol (DAG) functions as a second messenger signaling lipid, and is a product of the hydrolysis of the phospholipid PIP2 (phosphatidylinositolbisphosphate) by the enzyme phospholipase C (PLC) (a membrane-bound enzyme) that, through the same reaction, produces inositol trisphosphate (IP3). Although inositol trisphosphate (IP3) diffuses into the cytosol, DAG remains within the plasma membrane due to its hydrophobic properties. IP3 stimulates the release of calcium ions from the smooth endoplasmic reticulum, whereas DAG is a physiological activator of protein kinase C (PKC). The production of DAG in the membrane facilitates translocation of PKC from the cytosol to the plasma membrane.
Diacylglycerol mimicks the effects of the tumor-promoting compounds phorbol esters.

Application

18:1 DG may be used:
  • in the lipid-protein overlay screen assay with Arabidopsis Rho-like GTPases recombinant protein and Arabidopsis plasma membrane H+-ATPase
  • in the preparation of the Golgi-like liposomes
  • in giant unilamellar vesicles (GUVs) preparation

Biochem/physiol Actions

18:1 DG or 1,2-dioleoyl-sn-glycerol (DOG) fails to prevent the late fission events in the Golgi membrane. DOG is used in ferrihaem π–π tetramer aggregate preparation and β-haematin production.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800811C-10mg)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800811C-200mg)
30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800811C-25mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells:
Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration:
Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An improved protein lipid overlay assay for studying lipid-protein interactions
Han X, et al.
Plant methods, 16(1), 1-11 (2020)
Insights into the initial stages of lipid-mediated haemozoin nucleation
Kuter D, et al.
CrystEngComm, 18(27), 5177-5187 (2016)
Detecting phospholipase activity with the amphipathic lipid packing sensor motif of ArfGAP1
Quartino PY, et al.
Biochemical and Biophysical Research Communications, 505(1), 290-294 (2018)
Conical Lipids in Flat Bilayers Induce Packing Defects Similar to that Induced by Positive Curvature
Vamparys, L
Biophysical Journal, 104, 585-593 (2013)
Activation of ROP6 GTPase by phosphatidylglycerol in Arabidopsis
Han X, et al.
Frontiers in Plant Science, 9, 347-347 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service