Skip to Content
Merck
All Photos(1)

Key Documents

800800O

Avanti

08:0 DG

1,2-dioctanoyl-sn-glycerol, neat oil

Synonym(s):

DG(8:0/8:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O5
CAS Number:
Molecular Weight:
344.49
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 10 mg (with screw cap (800800O-10mg))
pkg of 1 × 25 mg (with screw cap (800800O-25mg))

manufacturer/tradename

Avanti Research - A Croda Brand 800800O

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C19H36O5/c1-3-5-7-9-11-13-18(21)23-16-17(15-20)24-19(22)14-12-10-8-6-4-2/h17,20H,3-16H2,1-2H3/t17-/m0/s1

InChI key

ZQBULZYTDGUSSK-KRWDZBQOSA-N

General description

08:0 DG (1,2-dioctanoyl-sn-glycerol), also referred as di-C8, is a short-chain cell-permeable diacyl glycerol.
In biochemical signaling, diacylglycerol (DAG) functions as a second messenger signaling lipid, and is a product of the hydrolysis of the phospholipid PIP2 (phosphatidylinositolbisphosphate) by the enzyme phospholipase C (PLC) (a membrane-bound enzyme) that, through the same reaction, produces inositol trisphosphate (IP3). Although inositol trisphosphate (IP3) diffuses into the cytosol, DAG remains within the plasma membrane due to its hydrophobic properties. IP3 stimulates the release of calcium ions from the smooth endoplasmic reticulum, whereas DAG is a physiological activator of protein kinase C (PKC). The production of DAG in the membrane facilitates translocation of PKC from the cytosol to the plasma membrane.
Diacylglycerol mimicks the effects of the tumor-promoting compounds phorbol esters.

Application

08:0 DG may be used:
  • to study its stimulatory effect on protein kinase C (PKC) in human cell lines
  • as a lipid cofactor in PKC activity assay
  • in vesicular-fusion protein Sec-18 binding studies

Biochem/physiol Actions

08:0 DG (1,2-dioctanoyl-sn-glycerol) mimics endogenous diacylglycerol andis a active phorbol compund. It activates protein kinase C (PKC). It binds to the epidermal growth factor (EGF) receptor similar to 4β-phorbol 12β-myristate 13α-acetate (PMA). DiC8 elevates the phosphoserine and phosphothreonine levels. DiC8 stimulates the release of superoxide in neutrophils. It downregulates nicotinic acetylcholine receptor α1 subunit 1 (nAChR1).

Packaging

5 mL PTFE Vial with Screw Cap (800800O-10mg)
5 mL PTFE Vial with Screw Cap (800800O-25mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells:
Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration:
Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rottlerin is a mitochondrial uncoupler that decreases cellular ATP
Soltoff SP
Test, 276(41), 37986-37992 (1986)
Regulation of human monocyte surface antigen expression. I. Up-modulation of Mo3e antigen expression on U-937 and HL-60 cells stimulated by pharmacologic activators of protein kinase C
Todd RF, et al.
Blood, 68(5), 1154-1161 (1986)
Phosphatidic acid induces conformational changes in Sec18 protomers that prevent SNARE priming
Starr ML, et al.
Test, 294(9), 3100-3116 (2019)
sn-1, 2-Dioctanoylglycerol. A cell-permeable diacylglycerol that mimics phorbol diester action on the epidermal growth factor receptor and mitogenesis.
Davis RJ, et al.
The Journal of biological chemistry, 260(3), 1562-1566 (1985)
Protein Kinase Calpha (PKCalpha) Is Resistant to Long Term Desensitization/Down-regulation by Prolonged Diacylglycerol Stimulation
Lum MA, et al.
The Journal of biological chemistry, 291(12), 6331-6346 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service