Skip to Content
Merck
All Photos(1)

Key Documents

790595P

Avanti

Upstate Lipid Blend

Synthetic Phospholipid Blend for PTEN, PDK1, and Akt1/PKBα Assay Kits, contains DOPC/DOPS (1:1, wt%), powder

Synonym(s):

Upstate Lipid Blend, Assay kits formerly supplied by Upstate Biotechnology.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 5 mg (790595P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790595P

lipid type

formulation
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Upstate lipid blend comprises DOPC:DOPS in the ratio 1:1wt%. 1,2-dioleoyl-sn-glycero-3-phosphocholine(DOPC) and 1,2-dioleoyl-sn-glycero-3-[phospho-l-serine] (DOPS) are crucial eukaryotic plasma membrane components.[1]

Application

Upstate lipid blend has been used in the generation for L-α-phosphatidylinositol (PI):DOPS:DOPC in phosphatidylinositol 4-kinase β (PI4Kβ) [2] and phosphoinositide 3-kinase β (PI3Kβ) activity studies.[3]

Biochem/physiol Actions

1,2-dioleoyl-sn-glycero-3-phosphocholine(DOPC) and 1,2-dioleoyl-sn-glycero-3-[phospho-l-serine] (DOPS) bilayers are stable and are used in generating multilamellar vesicles.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (790595P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular volumes of DOPC and DOPS in mixed bilayers of multilamellar vesicles
Murugova TN and Balgavy P
Physical Chemistry Chemical Physics, 16(34), 18211-18216 (2014)
Multistep compositional remodeling of supported lipid membranes by interfacially active phosphatidylinositol kinases
Tabaei SR, et al.
Analytical Chemistry, 88(10), 5042-5045 (2016)
Characterization of the interface of binary mixed DOPC: DOPS liposomes in water: The impact of charge condensation
Lutgebaucks C, et al.
J. Chem. Phys. , 146(4), 044701-044701 (2017)
Design and structural characterization of potent and selective inhibitors of phosphatidylinositol 4 kinase IIIbeta
Rutaganira FU, et al.
Journal of Medicinal Chemistry, 59(5), 1830-1839 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service