700071P
Avanti
24(R)-hydroxycholesterol
Avanti Research™ - A Croda Brand
Synonym(s):
5-cholestene-3β,24-diol ; 24-hydroxycholest-5-en-3-ol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Recommended Products
description
cholest-5-ene-3β,24(R)-diol
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (700071P-1mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−20°C
General description
24-hydroxycholesterol (24-OH) is an oxidized product of cholesterol. It is synthesized in the presence of enzyme 24S-hydroxylase (CYP46A1).
Biochem/physiol Actions
24(S)-hydroxycholesterol is preferred over 24(R)-hydroxycholesterol by human 7α-hydroxylase. However, the porcine enzyme is active towards both isomers. 24-hydroxycholesterol (24-OH) favors sirtuin 1 expression and reduces the tau protein phosphorylation.
Packaging
5 mL Amber Glass Screw Cap Vial (700071P-1mg)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gabriella Testa et al.
Redox biology, 17, 423-431 (2018-06-09)
It is now established that cholesterol oxidation products (oxysterols) are involved in several events underlying Alzheimer's disease (AD) pathogenesis. Of note, certain oxysterols cause neuron dysfunction and degeneration but, recently, some of them have been shown also to have neuroprotective
M Norlin et al.
Journal of lipid research, 41(10), 1629-1639 (2000-10-03)
(24S)-Hydroxycholesterol is formed from cholesterol in the brain and is important for cholesterol homeostasis in this organ. Elimination of (24S)-hydroxycholesterol has been suggested to occur in the liver but little is known about the metabolism of this oxysterol. In the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service