Skip to Content
Merck
All Photos(1)

Key Documents

541602P

Avanti

Soy Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

Soybean Polar Lipid Extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 g (541602P-1g)
pkg of 1 × 100 mg (541602P-100mg)
pkg of 5 × 100 mg (541602P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Polar Lipid Extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.
The Soy Extract Polar (SPE) comprises a mixture of phospholipids including phosphatidylcholine as a major component along with phosphatidylethanolamine phosphatidylinositol. It also has phosphatidic acid (along with other lipids each accounting to (6.9%). Soybean is the natural source of soy polar extract.

Application

Soy Extract Polar has been used:
  • as a component of wash and elution buffer for the purification of canonical transient receptor channels (TRPC) membrane protein
  • in the solubilization of ryanodine receptor channel protein
  • in the reconstitution of Ca2+-selective transient receptor potential vanilloid subfamily member 6 channel (TRPV6) on to nanodiscs

Biochem/physiol Actions

Soy Extract Polar (SPE) exists in hockey-puck-like structure and mimics real membranes. It is useful in the preparation of unilamellar liposomes and nanotube networks.

Packaging

5 mL Clear Glass Sealed Ampule (541602P-100mg)
5 mL Clear Glass Sealed Ampule (541602P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (541602P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of lipid mimetics on gating of ryanodine receptor
Willegems K and Efremov RG
Structure, 26(10), 1303-1313 (2018)
Opening of the human epithelial calcium channel TRPV6
McGoldrick LL, et al.
Nature, 553(7687), 233-237 (2018)
Membrane remodeling of giant vesicles in response to localized calcium ion gradients
Doosti BA
Journal of Visualized Experiments, e57789-e57789 (2018)
Electron cryo-microscopy structure of the canonical TRPC4 ion channel
Vinayagam D, et al.
eLife, 7, e36615-e36615 (2018)
Cryo-EM structures of the mammalian endo-lysosomal TRPML1 channel elucidate the combined regulation mechanism
Zhang S, et al.
Protein & cell, 8(11), 834-847 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service