Z680427
Duran® GL 45 connection system caps and accessories
GL 45 blue PP screw cap with 4 ports for HPLC system
Synonym(s):
1129812, Connection System Accessories, Duran Connection Caps, GL 45 Connection System
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
blue-gray polypropylene
manufacturer/tradename
DURAN® 1129812
parameter
-40-140 °C temp. limit
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Contains:
2x screw cap GL 45, 4 ports
2x set of 12x silicone seals (4 x 1.6 mm , 4 x 3.2 mm tubing seals, 4 x closure seals)
2x sets of 4x black M8 screw caps
The DURAN® HPLC bottle is available as a ready-made system. This comprises a special pressure resistant borosilicate glass bottle and a GL 45 screw cap with 4 ports. This enables 3 stations to be served at the same time from one reservoir. A filtered pressure equalisation set can be fitted to the fourth port. Complete system comprising: DURAN® pressure plus+ laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals.
2x screw cap GL 45, 4 ports
2x set of 12x silicone seals (4 x 1.6 mm , 4 x 3.2 mm tubing seals, 4 x closure seals)
2x sets of 4x black M8 screw caps
The DURAN® HPLC bottle is available as a ready-made system. This comprises a special pressure resistant borosilicate glass bottle and a GL 45 screw cap with 4 ports. This enables 3 stations to be served at the same time from one reservoir. A filtered pressure equalisation set can be fitted to the fourth port. Complete system comprising: DURAN® pressure plus+ laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals.
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Sterile pressure equalization is possible through use of membrane filter.
Features and Benefits
- Suitable for two different HPLC tubing outer diameters 1.6 or 3.2 mm
- Unused ports can be sealed with silicone closure / blanking seals
- Optional sterile pressure equalisation set (25 mm syringe filter 0.2 µm)
- Caps block solvent vapours VOCs, and keep your mobile phase clean. They improve lab health and safety, and the quality of your analysis
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service