Z676780
ART® low retention barrier tips
Tip design ART 1000XL, Reload insert
Synonym(s):
pipet tips, pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
Recommended Products
material
colorless
sterility
sterile
feature
barrier
Reload insert
Tip design ART 1000XL
packaging
pack of 4800 ea (50 inserts of 96 tips)
manufacturer/tradename
Molecular BioProducts 2179-05-RI
technique(s)
PCR: suitable
maximum volume
1 mL
Looking for similar products? Visit Product Comparison Guide
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service