Key Documents
Z303313
Quickfit® jacketed coil condenser
L 160 mm, joint: ST/NS 24/29
Synonym(s):
CX6/33/SC
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/29
manufacturer/tradename
Quickfit CX 6/33/SC
availability
not available in USA and Canada
not available in USA and Canada
L
160 mm
surface area
400 cm2
General description
Double surface condensers with a coil as the inner surface provides approximately twice the surface area of plain, double surface condensers of the same effective length.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)