Z303240
Quickfit® coil (Graham) condenser
L 160 mm, joint: ST/NS 19/26
Synonym(s):
C3/12/SC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS19/26
manufacturer/tradename
Quickfit C3/12/SC
availability
not available in USA and Canada
L
160 mm
surface area
200 cm2
Looking for similar products? Visit Product Comparison Guide
General description
The surface area in these coil condensers is considerably greater than that in a Liebig condenser of similar external dimensions. They have a wide vapour passage and are free from external condensation of water vapour because the outer surface is air cooled. Extremely efficient when used for reflux.
Supplied with detachable plastic screwthread connectors which allow 9 mm flexible tubing to be fitted easily and safely.
Supplied with detachable plastic screwthread connectors which allow 9 mm flexible tubing to be fitted easily and safely.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service