Skip to Content
Merck
All Photos(1)

Key Documents

Z278513

Wilmad® coaxial insert

stem L 50 mm, reference capacity 60 μL, sample capacity 530 μL

Synonym(s):

Wilmad nmr tubes, nmr sample tubes, nmr tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121705
NACRES:
SB.32

packaging

pack of 1 ea

manufacturer/tradename

Wilmad WGS-5BL

stem L

50 mm

Looking for similar products? Visit Product Comparison Guide

General description

Precision, all-glass construction virtually eliminates sidebands. Superior to PTFE inserts in variable-temperature experiments. Upper section of insert fits snugly into the outer NMR tube; 2 mm O.D. insert stem is held straight within 0.013 mm. Use inserts with 5 mm O.D. × 7 in. L thin-wall Precision NMR tubes only. Each insert supplied with a closing cap.
Coaxial inserts are applied in external referencing and locking, studying molecular interactions and determining magnetic susceptibility.

Outer tube must be ordered separately based on magnetic field strength.

Legal Information

Wilmad is a registered trademark of SP Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas D Werbeck et al.
Chembiochem : a European journal of chemical biology, 15(4), 543-548 (2014-02-13)
A variety of enzymes are activated by the binding of potassium ions. The potassium binding sites of these enzymes are very specific, but ammonium ions can often replace potassium ions in vitro because of their similar ionic radii. In these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service