Z108782
Stainless steel 304 syringe needle
gauge 16, L 6 in.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 1 ea
manufacturer/tradename
Popper 7046
L
6 in.
gauge
16
Looking for similar products? Visit Product Comparison Guide
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Standard Hub with 304 Stainless Steel Tubing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
PloS one, 12(12), e0189243-e0189243 (2017-12-15)
The insect repellent N,N-diethyl-3-methylbenzamide (DEET), is a multimodal compound that acts as a spatial repellent as well as an irritant (contact repellent), thus being perceived by the insect's olfactory and gustatory systems as an odorant and a tastant, respectively. Soon
Insect biochemistry and molecular biology, 113, 103224-103224 (2019-08-26)
In addition to its primary function as an insect repellent, DEET has many "off-label" properties, including a deterrent effect on the attraction of gravid female mosquitoes. DEET negatively affects oviposition sites. While deorphanizing odorant receptors (ORs) using the Xenopus oocyte
PloS one, 14(11), e0224810-e0224810 (2019-11-07)
Insect repellents are widely used as the first line of defense against mosquito bites and transmission of disease-causing agents. However, the cost of daily applications of even the most affordable and the gold standard of insect repellents, DEET, is still
Insect biochemistry and molecular biology, 113, 103213-103213 (2019-08-24)
Mosquitoes rely heavily on the olfactory system to find a host for a bloodmeal, plants for a source of energy and suitable sites for oviposition. Here, we examined a cluster of eight odorant receptors (ORs), which includes one OR, CquiOR1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service