WHA10539521
Whatman® protein saver card accessories
903 Dry rack with velcro, pack of 10 ea
Synonym(s):
Z699543, dry rack, protein saver card
All Photos(1)
About This Item
UNSPSC Code:
41111700
NACRES:
NB.79
Recommended Products
packaging
pack of 10 ea
manufacturer/tradename
Whatman 10539521
Whatman Article No. 28417176 (US reference)
General description
Whatman Dry Rak is designed to accommodate multiple collection forms at one time, properly air-drying the blood specimens in a suspended horizontal position. The Dry Rak is easily assembled and can be affixed to a wall or counter top with optional Velcro stickers.
Features and Benefits
- The Dry Rak is easily assembled and can be affixed to a wall or Counter® top with optional Velcro stickers.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Counter is a registered trademark of Cyanamid Company
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)