WHA10401314
Whatman® nitrocellulose NC20 filter discs 0.2 μm pore size
white, hydrophilic, 100 ea, 50 mm diam
Synonym(s):
Whatman filter, Z697206, cellulose membrane, membrane filter
About This Item
Recommended Products
product name
Whatman® nitrocellulose membrane filters, NC20 Membrane Circles, diam. 50 mm, pore size 0.2 μm
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 10401314
Whatman Article No. 28415006 (US reference)
diam.
50 mm
pore size
0.2 μm
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Narrow pore size distribution for improved surface capture and analysis.
- Low levels of extractables to ensure sample integrity.
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service