Skip to Content
Merck
All Photos(1)

Key Documents

W550132

Sigma-Aldrich

Trimethylamine solution

25 wt. % in propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
Beilstein:
956566
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.009
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

reg. compliance

FDA 21 CFR 117

concentration

25 wt. % in propylene glycol

refractive index

n20/D 1.356

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy

SMILES string

CN(C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3

InChI key

GETQZCLCWQTVFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Hillier
American journal of obstetrics and gynecology, 169(2 Pt 2), 455-459 (1993-08-01)
Bacterial vaginosis is characterized microbiologically by replacement of the Lactobacillus-predominant vaginal flora by Gardnerella vaginalis, Bacteroides species, Mobiluncus species, and genital mycoplasmas. A standardized, laboratory-based diagnostic test for bacterial vaginosis is desirable in those instances in which a microscope is
M Al-Waiz et al.
Clinical pharmacology and therapeutics, 42(5), 588-594 (1987-11-01)
Trimethylamine (TMA) and its N-oxide (TMAO) are normal components of human urine. They are present in the diet and also derived from the enterobacterial metabolism of precursors such as choline. Dietary TMA is almost entirely metabolized to and excreted as
Qian Li et al.
Current biology : CB, 23(1), 11-20 (2012-11-28)
Rodents use olfactory cues for species-specific behaviors. For example, mice emit odors to attract mates of the same species, but not competitors of closely related species. This implies rapid evolution of olfactory signaling, although odors and chemosensory receptors involved are
Smaranda Craciun et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(52), 21307-21312 (2012-11-16)
Choline and trimethylamine (TMA) are small molecules that play central roles in biological processes throughout all kingdoms of life. These ubiquitous metabolites are linked through a single biochemical transformation, the conversion of choline to TMA by anaerobic microorganisms. This metabolic
Flurin Cathomas et al.
Brain, behavior, and immunity, 50, 125-140 (2015-07-15)
The similarity between sickness behavior syndrome (SBS) in infection and autoimmune disorders and certain symptoms in major depressive disorder (MDD), and the high co-morbidity of autoimmune disorders and MDD, constitutes some of the major evidence for the immune-inflammation hypothesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service