Skip to Content
Merck
All Photos(1)

Documents

W526606

Sigma-Aldrich

(±)-Camphor

≥95.5%

Synonym(s):

1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
4513
Beilstein:
1907611
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Fragrance grade
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

vapor density

5.2 (vs air)

vapor pressure

4 mmHg ( 70 °C)

Assay

≥95.5%

expl. lim.

3.5 %

bp

204 °C (lit.)

mp

175-177 °C (lit.)

solubility

95% ethanol: soluble 1 mL/mL

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

camphor

Organoleptic

camphoraceous

SMILES string

[H][C@](CC1=O)(CC2)C(C)(C)[C@]12C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon P Skinner et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(29), 9022-9027 (2015-07-02)
The energy landscapes of proteins are highly complex and can be influenced by changes in physical and chemical conditions under which the protein is studied. The redox enzyme cytochrome P450cam undergoes a multistep catalytic cycle wherein two electrons are transferred
Christian Merten et al.
The journal of physical chemistry. A, 116(27), 7329-7336 (2012-06-06)
In the present work, the first observation of strong resonance Raman optical activity (RROA) involving more that one resonant electronic state is reported. The chiral transition metal complex bis-(trifluoroacetylcamphorato) copper(II), abbreviated Cu(tfc)(2), exhibits both resonance Raman (RR) and RROA spectra
Hiroyuki Kitahata et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 87(1), 010901-010901 (2013-02-16)
The coupling between deformation and motion in a self-propelled system has attracted broader interest. In the present study, we consider an elliptic camphor particle for investigating the effect of particle shape on spontaneous motion. It is concluded that the symmetric
Shigeki Yamamoto et al.
Journal of computational chemistry, 34(25), 2152-2158 (2013-07-05)
Induced resonance Raman optical activity (IRROA) proved to be a very sensitive method to detect molecular chirality. It is exhibited, for example, by complexes of lanthanides with chiral alcohols or ketones. So far, the phenomenon has not been understood at
Tamera J Corte et al.
American journal of respiratory and critical care medicine, 190(2), 208-217 (2014-06-18)
Pulmonary hypertension (PH) associated with fibrotic idiopathic interstitial pneumonia (IIP; idiopathic pulmonary fibrosis and nonspecific interstitial pneumonia) confers important additional morbidity and mortality. To evaluate the safety and clinical efficacy of the dual endothelin-1 receptor antagonist bosentan in this patient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service