Skip to Content
Merck
All Photos(1)

Key Documents

W357405

Sigma-Aldrich

4-Methylnonanoic acid

≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(CH3)CH2CH2CO2H
CAS Number:
Molecular Weight:
172.26
FEMA Number:
3574
EC Number:
Council of Europe no.:
11925
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.062
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

refractive index

n20/D 1.4355 (lit.)

bp

292-293 °C (lit.)

density

0.871 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty

SMILES string

CCCCCC(C)CCC(O)=O

InChI

1S/C10H20O2/c1-3-4-5-6-9(2)7-8-10(11)12/h9H,3-8H2,1-2H3,(H,11,12)

InChI key

WQTZCQIRCYSUBQ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca Hollenbach et al.
Molecules (Basel, Switzerland), 25(17) (2020-08-23)
Glycolipids are a class of biodegradable surfactants less harmful to the environment than petrochemically derived surfactants. Here we discuss interfacial properties, foam stability, characterized in terms of transient foam height, gas volume fraction and bubble diameter as well as texture
Hardy Z Castada et al.
Rapid communications in mass spectrometry : RCM, 31(19), 1641-1650 (2017-07-29)
4-Methyloctanoic acid, 4-ethyloctanoic acid, 4-methylnonanoic acid, and 3-methylindole are primary contributors to the distinctive aroma and flavor of lamb meat. The reactions of H The chemical ionization of pure standards of 4-ethyloctanoic acid, 4-methyloctanoic acid, 4-ethylnonanoic acid, and 3-methylindole was
Dorothee Eibler et al.
Journal of chromatography. A, 1505, 87-95 (2017-05-24)
Short chain 4-alkyl-branched fatty acids are character impact compounds of the flavor of sheep and goat milk and meat. Due to their methyl or ethyl branches these volatile fatty acids are chiral, and both enantiomers are characterized by different aroma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service