Skip to Content
Merck
All Photos(1)

Key Documents

W348007

Sigma-Aldrich

o-Cresol

≥99%

Synonym(s):

2-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
FEMA Number:
3480
Beilstein:
506917
EC Number:
Council of Europe no.:
618
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.027
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

vapor density

3.72 (vs air)

vapor pressure

0.3 mmHg ( 20 °C)
1 mmHg ( 38.2 °C)

Assay

≥99%

form

solid or liquid

autoignition temp.

1110 °F

expl. lim.

1.47 %, 148 °F

storage condition

under inert gas

refractive index

n20/D 1.541-1.550

pH

4.5 (25 g/L)

bp

191 °C (lit.)

mp

29-31 °C (lit.)

density

1.048 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

musty; phenolic

SMILES string

Cc1ccccc1O

InChI

1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3

InChI key

QWVGKYWNOKOFNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Cresol is one of the key volatile phenolic components of wine[1], beechwood creosote[2] and hardwood sawdust smoke.[3] It is also a flavor component of red meat[4] and aged Italian cheese.[5]

Application


  • Ultrahigh-performance liquid chromatography-tandem mass spectrometry quantitation of o-cresol in hydrolyzed human urine to assess toluene exposure.: This paper presents a method for quantifying o-cresol in human urine using advanced chromatography techniques, emphasizing its application in assessing exposure to toluene and other related compounds (Langlois et al., 2023).


Caution

Air sensitive

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antioxidant activity and characterization of volatile constituents of beechwood creosote.
Lee KG, et al.
Journal of Agricultural and Food Chemistry, 17(6), 1387-1393 (1969)
Volatile components of hardwood sawdust smoke. Components of phenolic fraction.
Lustre AO & Issenberg P.
Journal of Agricultural and Food Chemistry, 56(5), 1197-1202 (1991)
Volatile phenol determination in wine.
Etievant PX.
Journal of Agricultural and Food Chemistry, 85(9), 1580-1586 (2005)
Volatile Branched?chain Fatty Acids and Phenolic Compounds in Aged Italian Cheese Flavors.
Ha JK & Lindsay RC.
Journal of Food Science, 56(5), 1241-1247 (1991)
Simple phenol and phenolic compounds in food flavor
Maga JA & Katz I.
Critical Reviews in Food Science and Nutrition, 10(4), 323-372 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service