Skip to Content
Merck
All Photos(1)

Key Documents

W329355

Sigma-Aldrich

α-Angelica lactone

natural (US), ≥90%, sum of isomers

Synonym(s):

4-Hydroxy-3-pentenoic acid γ-lactone, 5-Methyl-2(3H)-furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6O2
CAS Number:
Molecular Weight:
98.10
FEMA Number:
3293
Beilstein:
108394
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
nutty; sweet; tobacco
grade:
Fragrance grade
Halal
Kosher
natural (US)
biological source:
botanical
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

botanical

Quality Level

grade

Fragrance grade
Halal
Kosher
natural (US)

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

Assay

≥90%

refractive index

n20/D 1.448 (lit.)

bp

55-56 °C/12 mmHg (lit.)

mp

13-17 °C (lit.)

density

1.092 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

nutty; sweet; tobacco

SMILES string

CC1=CCC(=O)O1

InChI

1S/C5H6O2/c1-4-2-3-5(6)7-4/h2H,3H2,1H3

InChI key

QOTQFLOTGBBMEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Recent regulatory changes in the US and EU have put the focus on fragrance safety and ingredient transparency, including the MoCRA, to be implemented within the next two years. Manufacturers and suppliers must comply with these changes to ensure they are meeting consumer demand and industry expectations for safety.

Recent regulatory changes in the US and EU have put the focus on fragrance safety and ingredient transparency, including the MoCRA, to be implemented within the next two years. Manufacturers and suppliers must comply with these changes to ensure they are meeting consumer demand and industry expectations for safety.

Recent regulatory changes in the US and EU have put the focus on fragrance safety and ingredient transparency, including the MoCRA, to be implemented within the next two years. Manufacturers and suppliers must comply with these changes to ensure they are meeting consumer demand and industry expectations for safety.

Recent regulatory changes in the US and EU have put the focus on fragrance safety and ingredient transparency, including the MoCRA, to be implemented within the next two years. Manufacturers and suppliers must comply with these changes to ensure they are meeting consumer demand and industry expectations for safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service