Skip to Content
Merck
All Photos(1)

Documents

W267600

Sigma-Aldrich

Methyl acetate

≥98%, FG

Synonym(s):

Methyl ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH3
CAS Number:
Molecular Weight:
74.08
FEMA Number:
2676
Beilstein:
1736662
EC Number:
Council of Europe no.:
213
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.023
NACRES:
NA.21

biological source

synthetic

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515
FDA 21 CFR 175.105

vapor density

2.55 (vs air)

vapor pressure

165 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

936 °F

expl. lim.

16 %

refractive index

n20/D 1.361 (lit.)

bp

57-58 °C (lit.)

mp

−98 °C (lit.)

density

0.934 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; ethereal; sweet

SMILES string

COC(C)=O

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl acetate has been identified as one of the volatile components in fresh grapefruit juice, green coffee and cantaloupe.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of ?ecological? post-harvest processing on the volatile fraction of coffee beans: I. Green coffee
Gonzalez-Rios O, et al.
J. Food Compos. Anal., 20(3), 289-296 (2007)
Identification of Additional Volatile Compounds from Cbntaloupe.
Senter S.
Journal of Food Science, 52(4), 1097-1098 (1987)
Analysis of volatile components in fresh grapefruit juice by purge and trap/gas chromatography.
Cadwallader KR & Xu Y
Journal of Agricultural and Food Chemistry, 42(3), 782-784 (1994)
Nicolas Salem et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 13(1), 140-151 (2010-04-20)
Studies have established the value of [(methyl)1-(11)C]-acetate ([(11)C]Act) combined with 2-deoxy-2[(18)F]fluoro-D-glucose (FDG) for detecting hepatocellular carcinoma (HCC) using positron emission tomography (PET). In this study, the metabolic fate of [(11)C]Act in HCC was characterized. Experiments with acetic acid [1-(14)C] sodium
Benjamin Bechem et al.
The Journal of organic chemistry, 75(5), 1795-1798 (2010-02-06)
5-Substituted-2-furan methanols 1a-c are subject to enantioselective carbonyl allylation, crotylation and tert-prenylation upon exposure to allyl acetate, alpha-methyl allyl acetate, or 1,1-dimethylallene in the presence of an ortho-cyclometalated iridium catalyst modified by (R)-Cl,MeO-BIPHEP, (R)-C3-TUNEPHOS, and (R)-C3-SEGPHOS, respectively. In the presence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service