Skip to Content
Merck
All Photos(2)

Key Documents

W256005

Sigma-Aldrich

trans-2-Hexen-1-al

≥95%, FCC, FG

Synonym(s):

trans-2-Hexenal, Leaf aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCHO
CAS Number:
Molecular Weight:
98.14
FEMA Number:
2560
Beilstein:
1699684
EC Number:
Council of Europe no.:
748
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.073
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

3.4 (vs air)

vapor pressure

10 mmHg ( 20 °C)

Assay

≥95%

impurities

<3% hexenoic acid

refractive index

n20/D 1.446 (lit.)

bp

47 °C/17 mmHg (lit.)

density

0.846 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; green; leafy; vegetable; fruity

storage temp.

2-8°C

SMILES string

CCC\C=C\C=O

InChI

1S/C6H10O/c1-2-3-4-5-6-7/h4-6H,2-3H2,1H3/b5-4+

InChI key

MBDOYVRWFFCFHM-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-2-Hexen-1-al is an important flavoring compound that occurs naturally in tomatoes, banana and black tea.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

100.9 °F - closed cup

Flash Point(C)

38.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavor of black tea.
Yamanishi T, et al.
Agricultural and Biological Chemistry, 30(8), 784-792 (1966)
(E)-2-Hexenal-Induced DNA Damage and Formation of Cyclic 1, N 2-(1, 3-Propano)-2 `-deoxyguanosine Adducts in Mammalian Cells.
Golzer P, et al.
Chemical Research in Toxicology, 9(7), 1207-1213 (1996)
Quantitative analysis of flavor parameters in six Florida tomato cultivars (Lycopersicon esculentum Mill).
Baldwin EA, et al.
Journal of Agricultural and Food Chemistry, 39(6), 1135-1140 (1991)
Kensuke Sakamaki et al.
Journal of agricultural and food chemistry, 59(12), 6667-6671 (2011-05-10)
A test apple beverage made up of apple juice (20%), high-fructose corn syrup (11.5%), citric acid (0.43%), trisodium citrate (0.02%), apple-odor flavor (0.1%), and ascorbic acid (0.02%) was stored at 40 °C and then analyzed for the change of odor
Min Jung Joo et al.
International journal of food microbiology, 153(3), 297-305 (2011-12-20)
Trans-2-hexenal, a naturally occurring plant volatile with antimicrobial capacity, was encapsulated into β-cyclodextrins (β-CDs), enzymatically modified starch, and shown effective to control main microorganisms causing food spoilage (Alternaria solani, Aspergillus niger, Botrytis cinerea, Colletotrichum acutatum, Penicillium sp). Loaded β-CDs were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service