Skip to Content
Merck
All Photos(1)

Key Documents

W245909

Sigma-Aldrich

Ethyl sorbate

≥97%, FG

Synonym(s):

Ethyl trans,trans-2,4-hexadienoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCO2C2H5
CAS Number:
Molecular Weight:
140.18
FEMA Number:
2459
Beilstein:
1362941
EC Number:
Council of Europe no.:
635
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.194
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.494 (lit.)

bp

82-83 °C/13 mmHg (lit.)

density

0.956 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; ethereal

SMILES string

CCOC(=O)\C=C\C=C\C

InChI

1S/C8H12O2/c1-3-5-6-7-8(9)10-4-2/h3,5-7H,4H2,1-2H3/b5-3+,7-6+

InChI key

OZZYKXXGCOLLLO-TWTPFVCWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl sorbate has been reported to be found in sparkling wine, when sorbic acid is used as a preservative.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wine constituents arising from sorbic acid addition, and identification of 2-ethoxyhexa-3, 5-diene as source of geranium-like off-odor
Crowell EA and JF Guymon
American Journal of Enology and Viticulture, 26(2), 97-102 (1975)
Sorbic acid as a preservative in sparkling wine. Its efficacy and adverse flavor effect associated with ethyl sorbate formation
De Rosa T, et al
American Journal of Enology and Viticulture, 34(2), 98-102 (1983)
Balasubramanian Narasimhan et al.
Bioorganic & medicinal chemistry letters, 17(21), 5836-5845 (2007-09-18)
A new series of 2,4-hexadienoic acid derivatives (S(1)-S(42)) has been synthesized and evaluated as antimicrobial agents against Staphylococcus aureus, Bacillus subtilis, Escherichia coli, Candida albicans, and Aspergillus niger. Quantitative structure-activity relationship (QSAR) investigation using Hansch analysis was applied to find

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service