Skip to Content
Merck
All Photos(1)

Key Documents

W229407

Sigma-Aldrich

Cinnamyl alcohol

≥98%, FG

Synonym(s):

3-Phenyl-2-propen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCH2OH
CAS Number:
Molecular Weight:
134.18
FEMA Number:
2294
Beilstein:
1903999
EC Number:
Council of Europe no.:
65
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.017
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

4.6 (vs air)

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

≥98%

bp

250 °C (lit.)

mp

30-33 °C (lit.)

density

1.044 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; balsamic; sweet

SMILES string

OC\C=C\c1ccccc1

InChI

1S/C9H10O/c10-8-4-7-9-5-2-1-3-6-9/h1-7,10H,8H2/b7-4+

InChI key

OOCCDEMITAIZTP-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cinnamyl alcohol is an α,β-unsaturated alcohol that can be used as a flavoring agent and a fragrance ingredient.

Application

Building block - Cinnamic alcohol is a building block for cinnamyl esters, via reaction with carboxylic acids or carboxylic acid derivatives. Some of the cinnamyl esters are used as fragrance components.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 601-602 (1997)
Cesar Rodriguez-Saona et al.
Annals of botany, 107(8), 1377-1390 (2011-04-19)
Studies of the effects of pollination on floral scent and bee visitation remain rare, particularly in agricultural crops. To fill this gap, the hypothesis that bee visitation to flowers decreases after pollination through reduced floral volatile emissions in highbush blueberries
Carmen Martínez et al.
Journal of molecular graphics & modelling, 28(2), 196-201 (2009-08-04)
The free radical reactivity of monolignols can be explained in terms of the Fukui function and the local hard and soft acids and bases (HSAB) principle to determine the potential linkages among them for reactions involving free radicals. Our results
Jin Zhang et al.
BMC genomics, 16, 181-181 (2015-04-19)
Heat shock proteins (Hsps) are molecular chaperones that are involved in many normal cellular processes and stress responses, and heat shock factors (Hsfs) are the transcriptional activators of Hsps. Hsfs and Hsps are widely coordinated in various biological processes. Although
Itai Chipinda et al.
Toxicology, 280(3), 135-143 (2010-12-18)
Consumer and medical products can contain leachable chemical allergens which can cause skin sensitization. Recent efforts have been directed at the development of non-animal based tests such as in vitro cell activation assays for the identification of skin sensitizers. Prohapten

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service