Skip to Content
Merck
All Photos(1)

Key Documents

W224847

Sigma-Aldrich

4-Carvomenthenol

≥95%, FCC, FG

Synonym(s):

4-Terpinenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2248
EC Number:
Council of Europe no.:
2229
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.072
NACRES:
NA.21
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.478

bp

88-90 °C

density

0.931 g/mL at 25

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; earthy; pepper

SMILES string

CC(C)C1(O)CCC(C)=CC1

InChI

1S/C10H18O/c1-8(2)10(11)6-4-9(3)5-7-10/h4,8,11H,5-7H2,1-3H3

InChI key

WRYLYDPHFGVWKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Anise, basil, black currant, cocoa, coriander, cranberry, grapefruit, lemon, lime, marjoram, peppermint oil, nutmeg and spearmint.

Application

  • 4-Carvomenthenol ameliorates the murine combined allergic rhinitis and asthma syndrome by inhibiting IL-13 and mucus production via p38MAPK/NF-κB signaling pathway axis.: Demonstrates the anti-inflammatory and therapeutic potentials of 4-Carvomenthenol in treating respiratory conditions, highlighting its mechanism of action at the molecular level (Bezerra Barros et al., 2020).

Biochem/physiol Actions

Taste at 30 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 59-59 (2005)
M Miyazawa et al.
Xenobiotica; the fate of foreign compounds in biological systems, 41(12), 1056-1062 (2011-11-08)
The in vitro metabolism of (-)-terpinen-4-ol was examined in human liver microsomes and recombinant enzymes. The biotransformation of (-)-terpinen-4-ol was investigated by gas chromatography-mass spectrometry. (-)-Terpinen-4-ol was found to be oxidized to (-)-(1S,2R,4R)-1,2-epoxy-p-menthan-4-ol, major metabolic product by human liver microsomal
Yong Jie Li et al.
Environmental science & technology, 44(14), 5483-5489 (2010-06-17)
We investigated the condensed-phase reactions of biogenic VOCs with C double bond C bonds (limonene, C(10)H(16), and terpineol, C(10)H(18)O) catalyzed by sulfuric acid by both bulk solution (BS) experiments and gas-particle (GP) experiments using a flow cell reactor. Product analysis
Antonella Casiraghi et al.
Pharmaceutical development and technology, 15(5), 545-552 (2009-10-22)
This work aimed to evaluate the effect induced by excipients conventionally used for topical dosage forms, namely isopropyl myristate (IPM) or oleic acid (OA) or polyethylene glycol 400 (PEG400) or Transcutol (TR), on the human skin permeability of terpinen-4-ol (T4OL)
Risa Haigou et al.
Journal of oleo science, 61(1), 35-43 (2011-12-23)
We examined the in vitro metabolism of (+)-terpinen-4-ol by human liver microsomes and recombinant enzymes. The biotransformation of (+)-terpinen-4-ol was investigated by gas chromatography-mass spectrometry (GC-MS). (+)-Terpinen-4-ol was found to be oxidized to (+)-(1R,2S,4S)-1,2-epoxy-p-menthan-4-ol, (+)-(1S,2R,4S)-1,2-epoxy-p-menthan-4-ol, and (4S)-p-menth-1-en-4,8-diol by human liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service