Skip to Content
Merck
All Photos(1)

Key Documents

W220302

Sigma-Aldrich

Butyl 4-hydroxybenzoate

≥99%

Synonym(s):

p-Hydroxybenzoic acid n-butyl ester, Butyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
194.23
FEMA Number:
2203
Beilstein:
1103741
EC Number:
Council of Europe no.:
525
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.754
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥99%

bp

156-157 °C/3.5 mmHg (lit.)

mp

67-70 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

faint

SMILES string

CCCCOC(=O)c1ccc(O)cc1

InChI

1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3

InChI key

QFOHBWFCKVYLES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Chen et al.
Journal of chromatography. A, 1591, 33-43 (2019-01-21)
A novel supramolecular solvent (SUPRAS) based on hexafluoroisopropanol (HFIP)/Brij-35 was proposed for liquid-phase microextraction (LPME) of parabens in water samples, pharmaceuticals and personal care products. Brij-35 is a cost-effective and non-toxic non-ionic surfactant, but it has a high cloud point
Guoyuan Wen et al.
The Journal of general virology, 96(Pt 6), 1219-1228 (2015-01-30)
Thermostable Newcastle disease virus (NDV) vaccines have been used widely to control Newcastle disease for village poultry flocks, due to their independence of cold chains for delivery and storage. To explore the potential use of thermostable NDV as a vaccine
Jordan Shaw et al.
Reproductive toxicology (Elmsford, N.Y.), 28(1), 26-31 (2009-06-06)
Parabens, a class of preservatives routinely added to cosmetics, pharmaceuticals, and foods, have estrogenic properties. Given that intrauterine implantation of fertilized ova in inseminated females can be disrupted by minute levels of exogenous estrogens, we assessed the impact of parabens
Caroline Stremnitzer et al.
The Journal of investigative dermatology, 135(7), 1790-1800 (2015-02-24)
Papain is commonly used in food, pharmaceutical, textile, and cosmetic industries and is known to induce occupational allergic asthma. We have previously shown that the papain-like cysteine protease Dermatophagoides pteronyssinus 1 from house dust mite exhibits percutaneous sensitization potential. We
Anna Viridén et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 77(1), 99-110 (2010-11-18)
This study investigates the effect of the chemical heterogeneity of hydroxypropyl methylcellulose (HPMC) on the release of model drug substances from hydrophilic matrix tablets. The hypothesis was that the release of drug substances could be influenced by possible interactions with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service