Skip to Content
Merck
All Photos(1)

Key Documents

W213000

Sigma-Aldrich

Benzaldehyde propylene glycol acetal, mixture of isomers

≥95%, FG

Synonym(s):

4-Methyl-2-phenyl-1,3-dioxolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2130
EC Number:
Council of Europe no.:
2226
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
6.032
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.509 (lit.)

bp

83-85 °C/4 mmHg (lit.)

density

1.065 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

bitter; woody; sweet

SMILES string

CC1COC(O1)c2ccccc2

InChI

1S/C10H12O2/c1-8-7-11-10(12-8)9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3

InChI key

CDIKGISJRLTLRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The product is an isomeric mixture of benzaldehyde propylene glycol acetal (Benzaldehyde PG acetal). It can be prepared from benzaldehyde and propylene glycol, via condensation reaction. Specific gravity of benzaldehyde PG acetal is 1.068 – 1.073 at 20°C. It has been identified as one of the volatile compound present in the artificial caramel and chocolate samples by non-equilibrated solid-phase microextraction/gas chromatography/mass spectrometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Headspace solid-phase microextraction analysis of artificial flavors.
Coleman WM and Dube MF.
Journal of the Science of Food and Agriculture, 85(15), 2645-2654 (2005)
Burdock GA.
Fenaroli's Handbook of Flavor Ingredients, Fifth Edition, 135-135 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service