Skip to Content
Merck
All Photos(2)

Key Documents

T90808

Sigma-Aldrich

L-Tyrosine methyl ester

98%

Synonym(s):

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid methyl ester, S-Tyrosine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)COOCH3
CAS Number:
Molecular Weight:
195.22
Beilstein:
2372626
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder with small lumps

optical activity

[α]20/D +26°, c = 2.4 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

134-136 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)[C@@H](N)Cc1ccc(O)cc1

InChI

1S/C10H13NO3/c1-14-10(13)9(11)6-7-2-4-8(12)5-3-7/h2-5,9,12H,6,11H2,1H3/t9-/m0/s1

InChI key

MWZPENIJLUWBSY-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshihiko Hanai et al.
Carbohydrate research, 498, 108171-108171 (2020-10-14)
Glycosylation, a part of the Maillard reaction, occurs non-enzymatically in food and biological processes. The selectivity of N-glycosylation was analyzed based on the reactivity of monosaccharides with aromatic amines, including aromatic amino acids, and the degree of molecular interaction (MI)
Xavier Belda et al.
Hormones and behavior, 126, 104865-104865 (2020-09-30)
Traumatic events have been proposed to be associated with hypo-activity of the hypothalamic-pituitary-adrenal (HPA) axis, but data in animal models exposed to severe stressors are controversial and have important methodological concerns. Individual differences in resting or stress levels of corticosterone
Federico Berti et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-12)
The synthesis of five hydroxycinnamoyl amides (HCAs) was accomplished and their identification and quantification in the green coffee bean samples of Coffeaarabica, Coffeacanephora, and Coffealiberica was performed. The HCAs p-coumaroyl-N-tyrosine 1b, caffeoyl-N-phenylalanine 2b, caffeoyl-N-tyrosine 3b, and p-coumaroyl-N-tryptophan 4b were characteristic
M J Barjavel et al.
European journal of pharmacology, 258(3), 173-178 (1994-06-13)
The effects of the methyl esters of L-tyrosine (L-Tyr-OMe) and L-tryptophan (L-Trp-OMe) on the analgesic action of trans-3,4-dichloro-N-methyl-N-[2-(1-pyrrolidin)cyclohexyl]-benzene acetamide methane sulfonate (U-50,488H), a kappa-opioid receptor agonist, were determined in male Swiss-Webster mice using the tail-flick test. Intraperitoneal injections of U-50,488H
Separation of 125I-labelled prostaglandin E2-tyrosine methyl ester by reversed-phase high-performance liquid chromatography.
I Mucha et al.
Journal of chromatography, 438(1), 111-116 (1988-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service