Skip to Content
Merck
All Photos(1)

Documents

T66206

Sigma-Aldrich

1,2,6-Hexanetriol

96%

Synonym(s):

1,2,6-Trihydroxyhexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)4CH(OH)CH2OH
CAS Number:
Molecular Weight:
134.17
Beilstein:
1304479
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.63 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

96%

refractive index

n20/D 1.476 (lit.)

bp

178 °C/5 mmHg (lit.)

density

1.109 g/mL at 25 °C (lit.)

SMILES string

OCCCCC(O)CO

InChI

1S/C6H14O3/c7-4-2-1-3-6(9)5-8/h6-9H,1-5H2

InChI key

ZWVMLYRJXORSEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,6-Hexanetriol has been used as a precursor to synthesize a trivalent linker containing three thiol moieties (triskelion) for the regioselective terminal activation of chitosan.
It can be used to prepare:
  • 1,6-Hexanediol (key polymer precursor to polyesters) via hydrodeoxygenation process in the presence of Rh–ReOx/SiO2 catalyst.
  • Tetrahydropyran-2-methanol, a six-membered oxide ring, by reacting with ethylene carbonate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

383.0 °F - closed cup

Flash Point(C)

195 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclic ethers made by pyrolysis of carbonate esters
Pattison DB
Journal of the American Chemical Society, 79(13), 3455-3456 (1957)
From 5-hydroxymethylfurfural (HMF) to polymer precursors: catalyst screening studies on the conversion of 1, 2, 6-hexanetriol to 1, 6-hexanediol
Buntara T, et al.
Topics in Catalysis, 55(7-10), 612-619 (2012)
Sayyeda Marziya Hasan et al.
Polymers, 12(10) (2020-10-11)
Shape memory polymer foams have been used in a wide range of medical applications, including, but not limited to, vessel occlusion and aneurysm treatment. This unique polymer system has been proven to shape-fill a void, which makes it useful for
Regioselective chitosan end-group activation: the triskelion approach
Pickenhahn VD, et al.
Royal Society of Chemistry Advances, 7(30), 18628-18638 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service