Skip to Content
Merck
All Photos(1)

Key Documents

T54801

Sigma-Aldrich

2,2,2-Trichloroethanol

ReagentPlus®, ≥99%

Synonym(s):

Trichloroethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCH2OH
CAS Number:
Molecular Weight:
149.40
Beilstein:
1697495
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.16 (vs air)

vapor pressure

1 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

refractive index

n20/D 1.49 (lit.)

bp

152-154 °C (lit.)

mp

17.8 °C (lit.)

density

1.56 g/mL at 20 °C (lit.)

SMILES string

OCC(Cl)(Cl)Cl

InChI

1S/C2H3Cl3O/c3-2(4,5)1-6/h6H,1H2

InChI key

KPWDGTGXUYRARH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

The 2,2,2-Trichloroethyl (TCE) moiety of 2,2,2-trichloroethanol is mainly used as a protecting group for acids, acetals and also phosphoryl groups of nucleotides. It can be easily cleaved by zinc reduction.
Some of its other application include:
  • Conversion of per-O-acetylated sugars to per-O-acetylated trichloroethyl derivatives.
  • Preparation of TCE ester of chlorosulfuric acid.
  • Preparation of 2,2,2-trichloroethyl acetimidate hydrochloride.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

e-EROS Encyclopedia of Reagents for Organic Synthesis null
Preparation of N1?Phenylacetamidine 4?Bromobenzoate Using 2, 2, 2?Trichloroethyl Acetimidate Hydrochloride.
Wei L & Caron S.
Organic Syntheses, 174-181 null
2, 2, 2-Trichloroethyl chloroformate: a general reagent for demethylation of tertiary methylamines.
Montzka TA, et al.
Tetrahedron Letters, 15(14), 1325-1327 (1974)
Sophie Scheidecker et al.
Human mutation, 40(10), 1826-1840 (2019-05-23)
Mutations in genes encoding aminoacyl-tRNA synthetases have been reported in several neurological disorders. KARS is a dual localized lysyl-tRNA synthetase and its cytosolic isoform belongs to the multiple aminoacyl-tRNA synthetase complex (MSC). Biallelic mutations in the KARS gene were described
Anand Chopra et al.
Scientific reports, 9(1), 13923-13923 (2019-09-29)
The incorporation of 2,2,2-trichloroethanol in polyacrylamide gels allows for fluorescent visualization of proteins following electrophoresis. Ultraviolet-light exposure, in the presence of this trichlorinated compound, results in a covalent modification of the tryptophan indole ring that shifts the fluorescent emission into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service